Skip to Content
Merck
All Photos(1)

Key Documents

45333

Supelco

Azinphos-methyl

PESTANAL®, analytical standard

Synonym(s):

Guthion®, Methyltriazotin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N3O3PS2
CAS Number:
Molecular Weight:
317.32
Beilstein:
280476
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(OC)SCN1N=Nc2ccccc2C1=O

InChI

1S/C10H12N3O3PS2/c1-15-17(18,16-2)19-7-13-10(14)8-5-3-4-6-9(8)11-12-13/h3-6H,7H2,1-2H3

InChI key

CJJOSEISRRTUQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Guthion is a registered trademark of Makhteshim Chemical Works Ltd.
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jianshe Tang et al.
Journal of immunoassay & immunochemistry, 29(4), 356-369 (2008-09-30)
Organophosphorus Pesticides (OPPs) are a group of artificially synthesized substances used in farms to control pests and to enhance agricultural production. Although these compounds show preferential toxicity to insects, they are also toxic to humans and mammals by the same
Silvia Buono et al.
Ecotoxicology (London, England), 21(3), 688-697 (2011-11-22)
The application of many current-use pesticides has increased after the disuse of persistent, bioaccumulative or toxic ones as DDT or chlordane. Many of the used pesticides are considered less dangerous towards the environment for their physico-chemical properties. This study investigated
Luis Claudio Cacciatore et al.
Chemosphere, 88(4), 450-458 (2012-03-23)
In this study, the cholinesterase (ChE) and carboxylesterase (CES) activities present in whole organism homogenates from Planorbarius corneus and their in vitro sensitivity to organophosphorous (OP) pesticides were studied. Firstly, a characterization of ChE and CES activities using different substrates
Jimena Soleño et al.
Pest management science, 68(11), 1451-1457 (2012-06-08)
Resistance to insecticides has been related to application history, genetic factors of the pest and the dynamic within the treated area. The aim of this study was to assess the geographic variation in azinphos-methyl response and the role of esterase
Alan L Knight
Pest management science, 66(8), 865-874 (2010-07-07)
Codling moth, Cydia pomonella (L.), has been intensely managed with the organophosphate insecticide azinphos-methyl for 50 years, and populations have developed resistance. New management programs have been developed and implemented that rely more heavily on other classes of insecticides. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service