Skip to Content
Merck
All Photos(2)

Documents

45477

Supelco

Ethion

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H22O4P2S4
CAS Number:
Molecular Weight:
384.48
Beilstein:
1804530
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)SCSP(=S)(OCC)OCC

InChI

1S/C9H22O4P2S4/c1-5-10-14(16,11-6-2)18-9-19-15(17,12-7-3)13-8-4/h5-9H2,1-4H3

InChI key

RIZMRRKBZQXFOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

G K Bhatti et al.
Environmental toxicology and pharmacology, 31(3), 378-386 (2011-07-27)
Organophosphate (OP) insecticides have been reported to induce oxidative stress due to lipid peroxidation and alteration in defense mechanisms. It is known that calcium content in erythrocytes plays a very important in normal physiology of cells. Erythrocytes are a very
Neera Singh et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 44(3), 214-219 (2009-03-13)
The sorption of carbaryl (1-naphthyl methyl carbamate) and ethion [O,O,O',O'-tetraethyl S,S'-methylene bis(phosphorodithioate)] was studied in whole soils as well as after treatment of soil with 2% hydrofluoric acid (HF) to remove paramagnetic materials and to oxidize most forms of labile
M W Bianchi et al.
Veterinary parasitology, 112(1-2), 75-89 (2003-02-13)
Boophilus microplus, even in the absence of babesiosis, is a major disease of the cattle in New Caledonia where the particular farming system associates continental European breeds and a tropical climate tempered by the Pacific Ocean. In order to have
I da Silva Vaz Jnr et al.
Insect molecular biology, 13(3), 329-335 (2004-05-26)
The ticks Haemaphysalis longicornis and Rhipicephalus appendiculatus are important parasites worldwide. The current method for control of cattle ticks involves the use of chemicals. Nevertheless, parasite resistance is an ever increasing global problem. Glutathione S-transferases (GSTs) play a central role
O S Anziani et al.
Veterinary parasitology, 91(1-2), 147-151 (2000-07-13)
A field study was conducted in central Argentina to evaluate the efficacy of ear tags containing 36% ethion against pyrethroid resistant populations of Haematobia irritans on grazing dairy cattle. The treated group consisted of 45 milking Holstein cows which received

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service