Skip to Content
Merck
All Photos(2)

Documents

46302

Supelco

Mefenpyr-diethyl

PESTANAL®, analytical standard

Synonym(s):

1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18Cl2N2O4
CAS Number:
Molecular Weight:
373.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)C1=NN(c2ccc(Cl)cc2Cl)C(C)(C1)C(=O)OCC

InChI

1S/C16H18Cl2N2O4/c1-4-23-14(21)12-9-16(3,15(22)24-5-2)20(19-12)13-7-6-10(17)8-11(13)18/h6-8H,4-5,9H2,1-3H3

InChI key

OPGCOAPTHCZZIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mefenpyr-diethyl is a herbicide safener, used for post emergence control of selective grass weeds affecting crops such as wheat, rye and spring barley.

Application

Mefenpyr-diethyl may be used as an analytical reference standard for the quantification of the analyte in environmental samples using various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design of experiments and detailed uncertainty analysis to develop and validate a solid-phase microextraction/gas chromatography-mass spectrometry method for the simultaneous analysis of 16 pesticides in water
Passeport E, et al.
Journal of Chromatography A, 1217(33), 5317-5327 (2010)
Modern Crop Protection Compounds (2012)
A multi-residue method for characterization and determination of atmospheric pesticides measured at two French urban and rural sampling sites
Baraud L, et al.
Analytical and Bioanalytical Chemistry, 377(7-8), 1148-1152 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service