Skip to Content
Merck
All Photos(1)

Key Documents

33386

Supelco

Aldicarb

PESTANAL®, analytical standard

Synonym(s):

O-(Methylcarbamoyl)-2-methyl-2-(methylthio)propionaldehyd-oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O2S
CAS Number:
Molecular Weight:
190.26
Beilstein:
2209304
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(ON=CC(C)(C)SC)=O

InChI

1S/C7H14N2O2S/c1-7(2,12-4)5-9-11-6(10)8-3/h5H,1-4H3,(H,8,10)

InChI key

QGLZXHRNAYXIBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aldicarb is a white crystalline, N-methycarbamate ester of aliphatic oxime, which may be used as an insecticide in agriculture.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Richard C Raabe et al.
PloS one, 9(8), e105999-e105999 (2014-08-28)
Alcohol addiction is a widespread societal problem, for which there are few treatments. There are significant genetic and environmental influences on abuse liability, and understanding these factors will be important for the identification of susceptible individuals and the development of
J L Burgess et al.
Archives of internal medicine, 154(2), 221-224 (1994-01-24)
Aldicarb is the most potent of the commercially available carbamate pesticides and is an unusual source of acute human poisonings. We present the case of a 43-year-old man exposed to aldicarb who developed severe cholinergic symptoms and progressive weakness requiring
Toxicology of aldicarb.
R L Baron et al.
Reviews of environmental contamination and toxicology, 105, 1-70 (1988-01-01)
Pratima Pandey et al.
PLoS genetics, 17(2), e1009346-e1009346 (2021-02-02)
Ethanol is a widely used drug, excessive consumption of which could lead to medical conditions with diverse symptoms. Ethanol abuse causes dysfunction of memory, attention, speech and locomotion across species. Dopamine signaling plays an essential role in ethanol dependent behaviors
R L Baron
Environmental health perspectives, 102 Suppl 11, 23-27 (1994-12-01)
Aldicarb, the active ingredient in the insecticide TEMIK, was introduced to the agricultural community over 25 years ago. It has been registered worldwide to control a wide variety of insect, mite, and nematode pests in agriculture. The toxicological research database

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service