Skip to Content
Merck
All Photos(1)

Key Documents

45316

Supelco

Alachlor

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-2′,6′-diethyl-N-(methoxymethyl)acetanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20ClNO2
CAS Number:
Molecular Weight:
269.77
Beilstein:
2944476
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCc1cccc(CC)c1N(COC)C(=O)CCl

InChI

1S/C14H20ClNO2/c1-4-11-7-6-8-12(5-2)14(11)16(10-18-3)13(17)9-15/h6-8H,4-5,9-10H2,1-3H3

InChI key

XCSGPAVHZFQHGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanjun Xin et al.
Journal of hazardous materials, 192(3), 1812-1818 (2011-08-02)
Wormhole-shaped TiO(2)/Ti (WT) and nanotube-shaped TiO(2)/Ti (TNT) photoelectrodes were prepared by anodic oxidation method. The morphology and structure were characterized by scanning electron microscopy (SEM), X-ray diffraction (XRD) and X-ray photoelectron spectroscopy (XPS). It was found that both crystal types
Rita C Bicho et al.
Ecotoxicology (London, England), 22(1), 156-165 (2012-11-13)
Pesticide exposure has been related with thyroid disrupting effects in different vertebrate species. However, very little is known about the effects of these compounds in reptiles. In the Mediterranean area, lacertid lizards are the most abundant vertebrate group in agroecosystems
Jonathan D Byer et al.
Chemosphere, 82(8), 1155-1160 (2011-01-11)
Concerns regarding the impacts of pesticides on aquatic species and drinking water sources have increased demands on water quality monitoring programs; however the costs of sample analysis can be prohibitive. In this study we investigated enzyme-linked immunosorbent assay (ELISA) as
Fátima N Gil et al.
Environmental toxicology and chemistry, 30(11), 2506-2518 (2011-08-16)
Alachlor has been a commonly applied herbicide and is a substance of ecotoxicological concern. The present study aims to identify molecular biomarkers in the eukaryotic model Saccharomyces cerevisiae that can be used to predict potential cytotoxic effects of alachlor, while
Béatrice Lauga et al.
Environmental science and pollution research international, 20(2), 1089-1105 (2012-06-23)
Despite the implication of anaerobic soil communities in important functions related to C and N biogeochemical cycles, their responses to pesticides are rarely assessed. This study focused on the impact of alachlor, a chloroacetanilide herbicide, on two agricultural soils differing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service