Skip to Content
Merck
All Photos(1)

Key Documents

45321

Supelco

Ametryn

PESTANAL®, analytical standard

Synonym(s):

2-Ethylamino-4-isopropylamino-6-methylthio-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17N5S
CAS Number:
Molecular Weight:
227.33
Beilstein:
613099
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(NC(C)C)nc(SC)n1

InChI

1S/C9H17N5S/c1-5-10-7-12-8(11-6(2)3)14-9(13-7)15-4/h6H,5H2,1-4H3,(H2,10,11,12,13,14)

InChI key

RQVYBGPQFYCBGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jean Claudio Santos Costa et al.
Physical chemistry chemical physics : PCCP, 11(34), 7491-7498 (2009-08-20)
The behavior of Au nanorods and Ag nanocubes as analytical sensors was evaluated for three different classes of herbicides. The use of such anisotropic nanoparticles in surface-enhanced Raman scattering (SERS) experiments allows the one to obtain the spectrum of crystal
Bin Xu et al.
Journal of hazardous materials, 169(1-3), 586-592 (2009-05-09)
The chemical oxidation of the herbicide ametryn was investigated by aqueous chlorination between pH 4 and 10 at a temperature of 25 degrees C. Ametryn was found to react very rapidly with aqueous chlorine. The reaction kinetics can be well
Analu Egydio Jacomini et al.
Archives of environmental contamination and toxicology, 60(3), 452-461 (2010-06-23)
São Paulo state, Brazil, is one of the main areas of sugar cane agriculture in the world. Herbicides, in particular, ametryn, are extensively used in this extensive area, which implies that this herbicide is present in the environment and can
Marie E DeLorenzo et al.
Environmental toxicology, 28(7), 359-371 (2011-06-01)
Ecological risk assessments are, in part, based on results of toxicity tests conducted under standard exposure conditions. Global climate change will have a wide range of effects on estuarine habitats, including potentially increasing water temperature and salinity, which may alter
Djavanshir Djozan et al.
Journal of chromatography. A, 1216(12), 2211-2219 (2009-02-03)
A monolithic ametryn molecular-imprinted polymer based on a simple polymerization method was fabricated for use as new solid-phase microextraction (SPME) fiber, which can be coupled with GC and GC/MS for selective extraction and analysis of triazine herbicides. Methacrylic acid (MAA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service