Skip to Content
Merck
All Photos(1)

Key Documents

37894

Supelco

Imidacloprid

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10ClN5O2
CAS Number:
Molecular Weight:
255.66
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

140-146 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

[O-][N+](=O)NC1=NCCN1Cc2ccc(Cl)nc2

InChI

1S/C9H10ClN5O2/c10-8-2-1-7(5-12-8)6-14-4-3-11-9(14)13-15(16)17/h1-2,5H,3-4,6H2,(H,11,13)

InChI key

YWTYJOPNNQFBPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Imidacloprid is a systemic chloronicotinyl insecticide, which can be effectively used in agriculture to control pests like aphids, scale insects, whiteflies, leafhoppers, some coleoptera and some lepidoptera species.

Find more information here: Neonicotinoids

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quancai Sun et al.
Journal of agricultural and food chemistry, 64(49), 9293-9306 (2016-12-15)
Imidacloprid, a neonicotinoid insecticide widely used in agriculture worldwide, has been reported to promote adipogenesis and cause insulin resistance in vitro. The purpose of the current study was to determine the effects of imidacloprid and its interaction with dietary fat
Alison R Holt et al.
The Science of the total environment, 573, 1422-1429 (2016-08-20)
Debate about how sustainable intensification and multifunctionality might be implemented continues, but there remains little understanding as to what extent they are achievable in arable landscapes. Policies that influence agronomic decisions are rarely made with an appreciation of the trade-offs
DDT resistance in Drosophila correlates with Cyp6g1 over-expression and confers cross-resistance to the neonicotinoid imidacloprid
Decourtye A, et al.
Molecular Genetics and Genomics, 266, 556-563 (2001)
Lin-Rui Gao et al.
Journal of agricultural and food chemistry, 64(47), 9078-9088 (2016-10-30)
As a neonicotinoid pesticide, imidacloprid is widely used to control sucking insects on agricultural planting and fleas on domestic animals. However, the extent to which imidacloprid exposure has an influence on cardiogensis in early embryogenesis is still poorly understood. In
Abhishek Mandal et al.
The Science of the total environment, 577, 376-385 (2016-11-08)
Agri-wastes biochars viz. eucalyptus bark (EBBC), corn cob (CCBC), bamboo chips (BCBC), rice husk (RHBC) and rice straw (RSBC) and acid treated RSBC (T-RSBC), were characterized for their physico-chemical properties and sorption behaviour of atrazine and imidacloprid was studied. Kinetics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service