Skip to Content
Merck
All Photos(1)

Key Documents

43107

Sigma-Aldrich

Cyanocobalamin

tested according to Ph. Eur.

Synonym(s):

Vitamin B12, α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H88CoN14O14P
CAS Number:
Molecular Weight:
1355.37
Beilstein:
4122889
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

fermentation/recombinant

Quality Level

Agency

USP/NF
tested according to Ph. Eur.

Assay

96.0-102.0% dry basis

form

powder

color

dark red

solubility

water: soluble 1 gm in 80 ml
acetone: insoluble
alcohol: soluble
chloroform: insoluble
diethyl ether: insoluble

application(s)

cell analysis

storage temp.

2-8°C

SMILES string

C/C(C1=N[C@]([C@]([C@H](CC(N)=O)[C@@]/2(C)CCC(NC[C@@H](OP([O-])(O[C@H]3[C@@H](O)[C@@H](N4C=[N]5C6=C4C=C(C)C(C)=C6)O[C@@H]3CO)=O)C)=O)([H])N([Co+]5C#N)C2=C(C)/C([C@@H](CCC(N)=O)C/7(C)C)=NC7=C/8)(C)[C@@](C)(CC(N)=O)[C@@H]1CCC(N)=O)=C9N=C8[C@@H](CCC(N)=O)[C@

InChI

1S/C62H90N13O14P.CN.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57?,59-,60+,61+,62+;;/m1../s1

InChI key

RMRCNWBMXRMIRW-QJRSUKKJSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Cyanocobalamin, also referred to as Vitamin B12, is a water soluble vitamin.[1]

Application

Cyanocobalamin was added in the assay buffer during the analysis step of ELISA for ensuring that all transcobalamin was saturated with cobalamin before analysis.[2] Cyanocobalamin was also used for examining its impact on the expression of mPer1 and mPer2 in vivo.[3]

Biochem/physiol Actions

Cyanocobalamin plays a role in functioning of brain and nervous system. It is also involved in hematopoiesis. Vitamin B12 deficiency leads to megaloblastic anemia. It is also associated with irreversible nervous system breakdown leading to spinal cord degeneration.[1]
Cyanocobalamin refers to a group of chemically-related cobalt containing molecules involved in cell processes such as DNA synthesis, fatty acid synthesis, energy production and regulation. Cyanocobalamin along with other B vitamins thiamine (B1) and pyridoxine (B6) blocks the thermal hyperalgesia in rats with neuropathic pain caused by spinal ganglia compression (CCD) or loose ligation of the sciatic nerve (CCI).[4] It can stimulate the anaerobic biodegradation of chloroform.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anti-nociceptive and anti-inflammatory effects of cyanocobalamin (vitamin B12) against acute and chronic pain and inflammation in mice
Hosseinzadeh H, et al.
Arzneimittelforschung, 62(07), 324-329 (2012)
Zheng-Bei Wang et al.
Pain, 114(1-2), 266-277 (2005-03-01)
Neuropathic pain after nerve injury is severe and intractable, and current drugs and nondrug therapies offer substantial pain relief to no more than half of affected patients. The present study investigated the analgesic roles of the B vitamins thiamine (B1)
E Nexo et al.
Clinical chemistry, 46(10), 1643-1649 (2000-10-06)
Transcobalamin is essential for the cellular internalization of cobalamin. Methods to quantify the unsaturated protein are available, but few attempts have been made to develop methods to quantify the sum of unsaturated and cobalamin saturated transcobalamin. gamma-Globulins from two polyclonal
Krista Kaasik et al.
Nature, 430(6998), 467-471 (2004-07-23)
The circadian clock is the central timing system that controls numerous physiological processes. In mammals, one such process is haem biosynthesis, which the clock controls through regulation of the rate-limiting enzyme aminolevulinate synthase 1 (Alas1). Several members of the core
Bamini Gopinath et al.
The American journal of clinical nutrition, 98(1), 129-135 (2013-05-03)
Epidemiologic evidence of a relation between serum total homocysteine (tHcy), vitamin B-12, and folate and age-related macular degeneration (AMD) is inconsistent and unresolved. In this cohort study, we aimed to investigate associations between intakes and serum concentrations of folate and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service