Skip to Content
Merck
All Photos(3)

Documents

H8017

Sigma-Aldrich

Hydroxocobalamin acetate

vitamin B12 analog

Synonym(s):

Vitamin B12a acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H91CoN13O16P
CAS Number:
Molecular Weight:
1388.39
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058004
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic (organic)

Assay

≥95% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

red to brown

storage temp.

2-8°C

SMILES string

CC(CNC(=O)CC[C@]1(C)[C@@H](CC(N)=O)[C@H]2N=C1C(C)=C3N=C(C=C4N=C(C(C)=C5[C@@H](CCC(N)=O)[C@](C)(CC(N)=O)[C@@]2(C)N5[Co]OC(C)=O)[C@@](C)(CC(N)=O)[C@@H]4CCC(N)=O)C(C)(C)[C@@H]3CCC(N)=O)OP([O-])(=O)O[C@H]6[C@@H](O)[C@H](O[C@@H]6CO)n7cnc8cc(C)c(C)cc78

InChI

1S/C62H90N13O14P.C2H4O2.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2(3)4;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);1H3,(H,3,4);/q;;+2/p-3/t31?,34-,35-,36-,37+,41-,52-,53-,56-,57+,59-,60+,61+,62+;;/m1../s1

InChI key

QHSPIHUMLGFPKX-WYVZQNDMSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Hydroxocobalamin is an analog of vitamin B12 with OH as an alkyl group. Vitamin B12 cobalamin refers to a group of chemically-related cobalt containing molecules.

Biochem/physiol Actions

Vitamin B12 cobalamin is involved in DNA synthesis and fatty acid synthesis. It also plays a vital role as a coenzyme in the conversion of mitochondrial methylmalonyl co-enzyme A to succinyl co-enzyme A. Vitamin B12 cobalamin is also involved in neurotransmitter synthesis and erythropoiesis. It also functions in the synthesis and maintenance of myelin sheath. Hydroxocobalamin (vitamin B12a, OHCbl) and cyanocobalamin (CNCbl) are storage and delivery forms.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photodegradation of cobalamins in aqueous solutions and in human blood
Juzeniene A and Nizauskaite Z
Journal of Photochemistry and Photobiology. B, Biology, 122(21), 7-14 (2013)
Hydroxocobalamin association during cell culture results in pink therapeutic proteins
MAbs, 5(6), 974-981 (2013)
The effect of vitamin B12 deprivation on the enzymes of fatty acid synthesis
Frenkel E P, et al.
The Journal of Biological Chemistry, 248(21), 7540-7546 (1973)
Iron, vitamin B12 and folate
Moll R and Davis B
Medicine, 45(4), 198-203 (2017)
Markos Koutmos et al.
The Journal of biological chemistry, 286(34), 29780-29787 (2011-06-24)
An early step in the intracellular processing of vitamin B(12) involves CblC, which exhibits dual reactivity, catalyzing the reductive decyanation of cyanocobalamin (vitamin B(12)), and the dealkylation of alkylcobalamins (e.g. methylcobalamin; MeCbl). Insights into how the CblC scaffold supports this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service