Skip to Content
Merck
All Photos(2)

Documents

41865

Supelco

Fipronil-desulfinyl

PESTANAL®, analytical standard

Synonym(s):

5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethyl)-1H-pyrazole-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Cl2F6N4
CAS Number:
Molecular Weight:
389.08
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥94.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

composition

carbon content, 37.04%
hydrogen content, 1.04%
nitrogen content, 14.4%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

189-190 °C

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1c(c(nn1-c2c(Cl)cc(cc2Cl)C(F)(F)F)C#N)C(F)(F)F

InChI

1S/C12H4Cl2F6N4/c13-5-1-4(11(15,16)17)2-6(14)9(5)24-10(22)8(12(18,19)20)7(3-21)23-24/h1-2H,22H2

InChI key

JWKXVHLIRTVXLD-UHFFFAOYSA-N

General description

Find more information here fipronil

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hongping Chen et al.
Food chemistry, 246, 328-334 (2018-01-03)
A fast, sensitive and reliable method for the determination of fipronil and its metabolites in tea and chrysanthemum was developed using a modified QuEChERS technique and an ultra performance liquid chromatography Q-Exactive Orbitrap mass spectrometry. The mixture of adsorbents containing
Shi-Wei He et al.
Journal of chromatography. A, 1553, 16-24 (2018-04-25)
A novel hydrophilicity nano-titania coating modified magnetic graphene oxide (HTC-Mag-GO) has been synthesized. It has been evaluated in PRiME (process, robustness, improvements, matrix effects, ease of use) pass-through cleanup procedure for human blood prior to analysis of fipronil and its
Qiaozhen Guo et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(8), 1543-1552 (2018-05-19)
An easy-to-use method for determining the levels of fipronil and its metabolites (fipronil-desulfinyl, fipronil-sulfone and fipronil-sulfide) in chicken egg, muscle and cake was developed and validated using a modified quick, easy, cheap, effective, rugged and safe (QuEChERS) approach coupled with
Wenjian Lao
Water research, 188, 116510-116510 (2020-10-18)
Environmental risk assessment of complex chemical mixtures has increasingly been prioritized as a management goal, especially in the regulatory sector. Although fipronil and its three degradates (-sulfone, -sulfide and -desulfinyl) have been frequently quantified in waterways, little information is available
Andrew D Goff et al.
Aquatic toxicology (Amsterdam, Netherlands), 186, 96-104 (2017-03-11)
Endocrine disrupting compounds (EDCs) are now widely established to be present in the environment at concentrations capable of affecting wild organisms. Although many studies have been conducted in fish, less is known about effects in invertebrates such as decapod crustaceans.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service