Skip to Content
Merck
All Photos(2)

Key Documents

16785

Supelco

Fipronil

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Cl2F6N4OS
CAS Number:
Molecular Weight:
437.15
Beilstein:
8090115
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas (Argon)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

Nc1c(c(nn1-c2c(Cl)cc(cc2Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F

InChI

1S/C12H4Cl2F6N4OS/c13-5-1-4(11(15,16)17)2-6(14)8(5)24-10(22)9(7(3-21)23-24)26(25)12(18,19)20/h1-2H,22H2

InChI key

ZOCSXAVNDGMNBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Find more information here fipronil
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Özge Nur Kanat et al.
Neurotoxicity research, 37(1), 30-40 (2019-09-05)
Fipronil is a broad-spectrum insecticide belonging to the phenyl pyrazole chemical family. Fipronil disrupts gamma-aminobutyric acid (GABA) receptors in the central nervous system, thereby blocking GABA-gated chloride channels. Neurotoxic symptoms of fipronil poisoning in humans are typically associated with the
M K Rust et al.
Journal of medical entomology, 51(3), 638-643 (2014-06-06)
The monitoring of the susceptibility offleas to insecticides has typically been conducted by exposing adults on treated surfaces. Other methods such as topical applications of insecticides to adults and larval bioassays on treated rearing media have been developed. Unfortunately, baseline
Colin C D Tingle et al.
Reviews of environmental contamination and toxicology, 176, 1-66 (2002-11-22)
Fipronil is a highly effective, broad-spectrum insecticide with potential value for the control of a wide range of crop, public hygiene, amenity, and veterinary pests. It can generally be applied at low to very low dose rates to achieve effective
Virginia C Moser et al.
Toxicology and applied pharmacology, 282(2), 161-174 (2014-12-17)
There is increasing emphasis on the use of biomarkers of adverse outcomes in safety assessment and translational research. We evaluated serum biomarkers and targeted metabolite profiles after exposure to pesticides (permethrin, deltamethrin, imidacloprid, carbaryl, triadimefon, fipronil) with different neurotoxic actions.
Emily J Remnant et al.
Insect biochemistry and molecular biology, 54, 11-21 (2014-09-07)
Extensive use of older generation insecticides may result in pre-existing cross-resistance to new chemical classes acting at the same target site. Phenylpyrazole insecticides block inhibitory neurotransmission in insects via their action on ligand-gated chloride channels (LGCCs). Phenylpyrazoles are broad-spectrum insecticides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service