Skip to Content
Merck
All Photos(1)

Documents

46451

Supelco

Fipronil

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Cl2F6N4OS
CAS Number:
Molecular Weight:
437.15
Beilstein:
8090115
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

196-201 °C

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1c(c(nn1-c2c(Cl)cc(cc2Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F

InChI

1S/C12H4Cl2F6N4OS/c13-5-1-4(11(15,16)17)2-6(14)8(5)24-10(22)9(7(3-21)23-24)26(25)12(18,19)20/h1-2H,22H2

InChI key

ZOCSXAVNDGMNBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Find more information here fipronil

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grzegorz Buczkowski et al.
Scientific reports, 9(1), 19495-19495 (2019-12-22)
Invasive ants are major agricultural and urban pests and a significant concern in conservation areas. Despite long history of control and eradication efforts, invasive ants continue to spread around the globe driven by a multitude of synergistic factors. Lack of
Sunayana Saini et al.
Environmental monitoring and assessment, 186(1), 69-75 (2013-08-01)
Fipronil belongs to phenylpyrazole class of chemical compounds. Degradation of fipronil in sandy loam soil was investigated under field conditions by applying fipronil (Regent 5 % SC) at 50 (T 1) and 100 g a.i. ha(-1) (T 2) in field. Samples were
Thomas G Shelton
Journal of economic entomology, 106(5), 2160-2166 (2013-11-15)
A small-plot field trial was conducted to examine the area of influence of fipronil at incremental distances away from treated plots on the Harrison Experimental Forest near Saucier, MS. Small treated (water and fipronil) plots were surrounded by untreated wooden
Stephan M Carvalho et al.
Environmental toxicology and chemistry, 32(9), 2117-2124 (2013-05-29)
The present study was intended to evaluate the responses of enzymes in the honeybee Apis mellifera after exposure to deltamethrin, fipronil, and spinosad and their use as biomarkers. After determination of the median lethal doses (LD50), honeybees were exposed at
Cai Wang et al.
Journal of economic entomology, 106(6), 2630-2632 (2014-02-07)
Fipronil is a widely used insecticide for termite control. Although transfer of fipronil among termite cohorts has been investigated in previous studies, no study has yet focused on the influence of termite group size (density) on horizontal transfer. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service