Skip to Content
Merck
All Photos(2)

Key Documents

36171

Supelco

Molinate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NOS
CAS Number:
Molecular Weight:
187.30
Beilstein:
1239196
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCSC(=O)N1CCCCCC1

InChI

1S/C9H17NOS/c1-2-12-9(11)10-7-5-3-4-6-8-10/h2-8H2,1H3

InChI key

DEDOPGXGGQYYMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Molinate is a selective, preemergence, thiocarbamate herbicide, used for the control of grass weeds in rice cultivation.

Application

Molinate may be used as a reference standard for the determination of molinate in surface water samples by liquid chromatography-mass spectrometry with a particle beam interface using electron ionization and chemical ionization.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of UV spectrophotometric method for determining the herbicide molinate with and without alginate microparticles.
Paiva A M, et al.
Environmental Engineering and Management Journal, 14(2), 303-309 (2015)
Characterization of risks associated with the use of molinate.
Cochran R C, et al.
Regulatory Toxicology and Pharmacology, 25(2), 146-157 (1997)
Identification of pesticides by liquid chromatography--particle beam mass spectrometry using electron ionization and chemical ionization.
Aguilar C, et al.
Journal of Chromatography A, 805(1-2), 127-135 (1998)
Patrícia Correia et al.
Water research, 40(2), 331-340 (2005-12-29)
The effect of operating conditions during molinate degradation by the defined mixed bacterial culture DC, previously described as able to mineralize molinate, was evaluated in a batch reactor. Parameters such as the rate of molinate degradation, the dissolved organic carbon
Masanao Yamashita et al.
Regulatory toxicology and pharmacology : RTP, 70(1), 214-221 (2014-07-13)
To predict concentrations in humans of the herbicidal carbamate molinate, used exclusively in rice cultivation, a forward dosimetry approach was carried out using data from lowest-observed-adverse-effect-level doses orally administered to rats, wild type mice, and chimeric mice with humanized liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service