Skip to Content
Merck
All Photos(1)

Documents

36175

Supelco

Napropamide

PESTANAL®, analytical standard

Synonym(s):

N,N-Diethyl-2-(1-naphthyloxy)propanamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OCH(CH3)CON(CH2CH3)2
CAS Number:
Molecular Weight:
271.35
Beilstein:
6809160
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

73-78 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

CCN(CC)C(=O)C(C)Oc1cccc2ccccc12

InChI

1S/C17H21NO2/c1-4-18(5-2)17(19)13(3)20-16-12-8-10-14-9-6-7-11-15(14)16/h6-13H,4-5H2,1-3H3

InChI key

WXZVAROIGSFCFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Napropamide is a soil herbicide, which can be either sprayed on the surface or incorporated. It can be mainly used for weed control management.
Phenanthrene can be synthesized, via a multi-step synthesis process, using Haworth method involving naphthalene and succinic anhydride as the starting materials. It can undergo chlorination in acetic acid to form a mixture of cis- and trans-9,10-dichloro-9,10-dihydrophenanthrenes as the major products.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hepworth.DJ, et al.
Aromatic Chemistry, 13 (2002)
José P Da Silva et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 7(1), 69-75 (2008-01-03)
The photochemical behaviour of the herbicide napropamide is studied on cellulose and silica surfaces, using steady-state and laser-flash diffuse reflectance techniques. The results are used to probe how the reaction sites of the host matrices influence the photo-reactive pathways. Napropamide
Alfonso Salinas-Castillo et al.
Pest management science, 61(8), 816-820 (2005-04-20)
A new, simple, rapid and selective phosphorimetric method for determining napropamide is proposed which demonstrates the applicability of heavy-atom-induced room-temperature phosphorescence for analyzing pesticides in real samples. The phosphorescence signals are a consequence of intermolecular protection and are found exclusively
Fardin Sadegh-Zadeh et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 47(2), 144-151 (2012-01-19)
This study was carried out to determine the sorption-desorption, degradation and leaching of napropamide in selected Malaysian soils. The sorption capacities of the selected Malaysian soils for napropamide were the following in descending order: Linau > Teringkap > Gunung Berinchang
Pabitra Kumar Biswas et al.
Bulletin of environmental contamination and toxicology, 79(5), 566-569 (2007-10-10)
Napropamide is an amide group of herbicide, used as pre-emergence herbicide controlling in broad leaved weeds of tea, ground nut, citrus, etc. Napropamide 45 SC (Devrinol) was applied on tea bushes twice @ 1.125 and 2.250 g a.i./ha along with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service