About This Item
Recommended Products
grade
HPLC Plus
form
liquid
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
impurities
≤0.0003% non-volatile matter
≤1 ppb fluorescence (quinine) at 254 nm
≤1 ppb fluorescence (quinine) at 365 nm
refractive index
n20/D 1.34 (lit.)
pH
6-8 (25 °C)
bp
100 °C (lit.)
mp
0 °C (32 °F)
0 °C (lit.)
solubility
water: miscible
density
1.000 g/mL at 3.98 °C (lit.)
anion traces
chloride (Cl-): ≤0.1 mg/kg
fluoride (F-): ≤0.1 mg/kg
nitrate (NO3-): ≤0.1 mg/kg
sulfate (SO42-): ≤0.1 mg/kg
HPLC-gradient
≤1 mAU at 254 nm
≤5 mAU at 210 nm
application(s)
general analytical
SMILES string
O
InChI
1S/H2O/h1H2
InChI key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- To prepare the diluted hydrochloric acid solutions required for the preparation of standard solutions of amino acid.[1]
- In the sample extraction step during the gas chromatography mass spectrometric (GC-MS) analysis of free (FAA) and combined amino acids (CAA) in aerosol samples.[1]
- Preparation of tryptic digests from the standard protein samples and milk digests from milk samples.[2]
- Preparation of C60-aminopropylsilica.[2]
Preparation Note
Other Notes
- The article number 34877-4X2.5L-M will be discontinued. Please order the single bottle 34877-2.5L-M which is physically identical with the same exact specifications.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service