Skip to Content
Merck
All Photos(1)

Key Documents

33984

Supelco

Difloxacin hydrochloride

VETRANAL®, analytical standard

Synonym(s):

6-Fluoro-1-(4-fluorophenyl)-1,4-dihydro-7-(4-methylpiperazino)-4-oxo-3-quinolinecarboxylic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19F2N3O3 · HCl
CAS Number:
Molecular Weight:
435.85
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN1CCN(CC1)c2cc3N(C=C(C(O)=O)C(=O)c3cc2F)c4ccc(F)cc4

InChI

1S/C21H19F2N3O3.ClH/c1-24-6-8-25(9-7-24)19-11-18-15(10-17(19)23)20(27)16(21(28)29)12-26(18)14-4-2-13(22)3-5-14;/h2-5,10-12H,6-9H2,1H3,(H,28,29);1H

InChI key

JFMGBGLSDVIOHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gui Hong Fu et al.
Environmental toxicology and pharmacology, 31(2), 307-313 (2011-07-27)
Cytochrome P450s (CYPs) play a prominent role in drug metabolism and biotransformation which are distributed in liver of aquatic animals. However, limited information is available about CYP genes involved in drug metabolism in fish. In the present study, we explore
Irina Gerchman et al.
Veterinary microbiology, 131(3-4), 266-276 (2008-06-07)
Monitoring of susceptibility to antibiotics in field isolates of pathogenic avian mycoplasmas is important for appropriate choice of treatment. Our study compared in vitro susceptibility to enrofloxacin and difloxacin in recent (2005-2006) isolates of Mycoplasma gallisepticum and Mycoplasma synoviae from
K Abo-El-Sooud et al.
Veterinary journal (London, England : 1997), 179(2), 282-286 (2008-07-08)
Single-dose disposition kinetics of difloxacin (5mg/kg bodyweight) were determined in clinically normal male dromedary camels (n=6) following intravenous (IV) and intramuscular (IM) administration. Difloxacin concentrations were determined by high performance liquid chromatography with fluorescence detection. The concentration-time data were analysed
Elias Gebru et al.
Veterinary microbiology, 154(3-4), 384-394 (2011-09-07)
The antibacterial activity, selection of Escherichia coli (E. coli) mutants and mechanisms of fluoroquinolone resistance were investigated by integrating the minimum inhibitory concentration (MIC), mutant prevention concentration (MPC) and in vitro dynamic model approaches. Difloxacin and orbifloxacin, for which the
H Z Ding et al.
Journal of veterinary pharmacology and therapeutics, 31(3), 200-204 (2008-05-13)
Pharmacokinetics of difloxacin, a fluoroquinolone antibiotic, was determined in pigs and broilers after intravenous (i.v.), intramuscular (i.m.), or oral (p.o.) administration at a single dose of five (pigs) or 10 mg/kg (broilers). Plasma concentration profiles were analyzed by a compartmental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service