Skip to Content
Merck
All Photos(1)

Key Documents

32372

Supelco

Cyantraniliprole

PESTANAL®, analytical standard

Synonym(s):

3-Bromo-1-(3-chloro-2-pyridyl)-4′-cyano-2′-methyl-6′-(methylcarbamoyl)pyrazole-5-carboxanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14BrClN6O2
CAS Number:
Molecular Weight:
473.71
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

O=C(C1=CC(Br)=NN1C2=C(Cl)C=CC=N2)NC3=C(C(NC)=O)C=C(C#N)C=C3C

InChI

1S/C19H14BrClN6O2/c1-10-6-11(9-22)7-12(18(28)23-2)16(10)25-19(29)14-8-15(20)26-27(14)17-13(21)4-3-5-24-17/h3-8H,1-2H3,(H,23,28)(H,25,29)

InChI key

DVBUIBGJRQBEDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyantraniliprole an anthranilic diamide, is a second generation ryanodine receptor insecticide. It is widely used to control sucking, chewing and Hemipteran insect pests.

Application

Cyantraniliprole may be used as a reference standard for the quantification of cyantraniliprole and its metabolites in samples of soil and vegetables by ultra-performance liquid chromatography/tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunmei Xu et al.
Journal of economic entomology, 110(1), 230-238 (2016-12-25)
To better understand the application prospect of cyantraniliprole against the Asian corn borer, Ostrinia furnacalis, a diet-incorporation bioassay was adopted to determine the toxicity of cyantraniliprole against the fourth instar of O. furnacalis. Moreover, other experiments were conducted to examine
First report of field resistance to cyantraniliprole, a new anthranilic diamide insecticide, on Bemisia tabaci MED in China.
Ran W, et al.
Journal of Integrative Agriculture, 17(1), 158-163 (2018)
Determination of cyantraniliprole and its major metabolite residues in vegetable and soil using ultra?performance liquid chromatography/tandem mass spectrometry.
Dong F, et al.
Biomedical Chromatography, 26(3), 377-383 (2012)
Jonghwa Lee et al.
Scientific reports, 9(1), 11648-11648 (2019-08-14)
The dissipation patterns of cyantraniliprole and its metabolite IN-J9Z38 were investigated using proso millet (Panicum miliaceum) under open-field conditions to establish a pre-harvest interval. A simple and sensitive analytical method was developed for analyzing residues using ultra-high performance liquid chromatography
Yayun Zuo et al.
Insect biochemistry and molecular biology, 89, 79-85 (2017-09-16)
Diamide insecticides selectively activate insect ryanodine receptors (RyRs), inducing uncontrolled release of calcium ions, and causing muscle contraction, paralysis and eventually death. The RyR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service