Skip to Content
Merck
All Photos(1)

Documents

33376

Supelco

β-HCH

PESTANAL®, analytical standard

Synonym(s):

β-1,2,3,4,5,6-Hexachlorocyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83
Beilstein:
1907338
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

≥300 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

Cl[C@H]1[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@@H](Cl)[C@@H]1Cl

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3+,4+,5-,6-

InChI key

JLYXXMFPNIAWKQ-CDRYSYESSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Hexachlorocyclohexane is an isomer of hexachlorocyclohexane. It is an organochlorine pesticide with carcinogenic effects on both humans and animals.

Application

β-Hexachlorocyclohexane may be used as a reference standard for the determination of β-hexachlorocyclohexane in marine sediments by pressurized liquid extraction (PLE) and stir-bar sorptive extraction (SBSE) coupled with thermal desorption and gas chromatography–triple quadrupole mass spectrometry (TD–GC–MS/MS QqQ).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Seasonal impacts on β-hexachlorocyclohexane concentration in soil solution.
Kalbitz K and Popp P
Environmental Pollution (Barking, Essex : 1987), 106(1), 139-141 (1999)
Validation of a GC?MS/MS method for simultaneous determination of 86 persistent organic pollutants in marine sediments by pressurized liquid extraction followed by stir bar sorptive extraction.
Camino-Sanchez FJ, et al.
Chemosphere, 84(7), 869-881 (2011)
Behaviour of α-, β-, γ-, and δ-hexachlorocyclohexane in the soil?plant system of a contaminated site.
Pereira RC, et al.
Environmental Pollution (Barking, Essex : 1987), 144(1), 210-217 (2006)
Miquel Porta et al.
Environment international, 44, 106-111 (2012-03-20)
Surveys of human exposure to environmental chemicals do not integrate the number of compounds detected per person and the concentration of each compound. This leaves untested relevant exposure scenarios, such as whether individuals with low concentrations of some compounds have
Tian Lin et al.
Marine pollution bulletin, 64(8), 1549-1555 (2012-06-16)
Forty-two surface sediment samples collected from the southern coastal East China Sea (ECS) were analyzed for organochlorine pesticides (OCPs). The results showed high concentrations of DDTs and parent DDT fractions in the nearshore area, suggesting that there is still a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service