Skip to Content
Merck
All Photos(1)

Documents

33699

Supelco

Enrofloxacin

VETRANAL®, analytical standard

Synonym(s):

Baytril

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22FN3O3
CAS Number:
Molecular Weight:
359.39
Beilstein:
5307824
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

clinical testing

format

neat

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

CCN1CCN(CC1)c2cc3N(C=C(C(O)=O)C(=O)c3cc2F)C4CC4

InChI

1S/C19H22FN3O3/c1-2-21-5-7-22(8-6-21)17-10-16-13(9-15(17)20)18(24)14(19(25)26)11-23(16)12-3-4-12/h9-12H,2-8H2,1H3,(H,25,26)

InChI key

SPFYMRJSYKOXGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: fluoroquinolone
Enrofloxacin is a broad-spectrum fluoroquinolone antibiotic extensively used in human and veterinary medicines. The drug acts by inhibiting bacterial DNA gyrase and DNA Topoisomerase IV (Topo IV), which are the vital enzymes responsible for DNA replication.

Application

Enrofloxacin may be used as an analytical reference standard for the determination of the analyte in bovine milk, pig plasma, goat milk, and food samples by various analytical techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple, rapid determination of enrofloxacin and ciprofloxacin in bovine milk and plasma by high-performance liquid chromatography with fluorescence detection
Idowu OR and Peggins JO
Journal of Pharmaceutical and Biomedical Analysis, 35(1), 143-153 (2004)
Determination of residual enrofloxacin in food samples by a sensitive method of chemiluminescence enzyme immunoassay
Yu F, et al.
Food Chemistry, 149, 71-75 (2014)
Determination of enrofloxacin and its metabolite ciprofloxacin in goat milk by high-performance liquid chromatography with diode-array detection: Optimization and validation
Cinquina AL, et al.
Journal of Chromatography A, 987(1-2), 221-226 (2003)
Determination of ciprofloxacin, enrofloxacin and flumequine in pig plasma samples by capillary isotachophoresis-capillary zone electrophoresis
Hernandez M, et al.
Journal of Chromatography. B, Biomedical Applications, 772(1), 163-172 (2002)
Jochen Springer et al.
Stroke, 45(12), 3675-3683 (2014-10-30)
Muscle wasting is a common complication accompanying stroke. Although it is known to impair poststroke recovery, the mechanisms of subacute catabolism after stroke have not been investigated in detail. The aim of this study is to investigate mechanisms of local

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service