Skip to Content
Merck
All Photos(1)

Key Documents

32468

Supelco

Nonylphenol solution

1000 μg/mL in acetone, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

1000 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)CCCCCCC(C)C

InChI

1S/C15H24O/c1-13(2)7-5-3-4-6-8-14-9-11-15(16)12-10-14/h9-13,16H,3-8H2,1-2H3

InChI key

JSFITYFUKSFPBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonylphenol belongs to the alkylphenol group of compounds.

Application

Nonylphenol may be used as an analytical reference standard for the determination of the analyte in:
  • Soils treated with sewage sludge by pressurized liquid extraction (PLE) followed by liquid chromatography-mass spectrometry (LC-MS).
  • Wastewater and sewage sludge by gas chromatography (GC) coupled to MS.
  • Wastewater treatment plant effluents and surface waters by solid phase extraction (SPE) followed by LC-MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Quantitative determination of octylphenol, nonylphenol, alkylphenol ethoxylates and alcohol ethoxylates by pressurized liquid extraction and liquid chromatography-mass spectrometry in soils treated with sewage sludges.
Andreu V, et al.
The Science of the Total Environment, 378(1-2), 124-129 (2007)
LC-MS-MS analysis and occurrence of octyl- and nonylphenol, their ethoxylates and their carboxylates in Belgian and Italian textile industry, waste water treatment plant effluents and surface waters.
Loos R, et al.J
Chemosphere, 66(4), 690-699 (2007)
Simultaneous determination of the endocrine disrupting compounds nonylphenol, nonylphenol ethoxylates, triclosan and bisphenol A in wastewater and sewage sludge by gas chromatography-mass spectrometry.
Gatidou G, et al.
Journal of Chromatography A, 1138(1-2), 32-41 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service