Skip to Content
Merck
All Photos(1)

Key Documents

384445

Sigma-Aldrich

4-Octylphenol

99%

Synonym(s):

4-Octylphenol, 4-n-Octylphenol, p-(n-Octyl)phenol, p-Octylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7C6H4OH
CAS Number:
Molecular Weight:
206.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

150 °C/4 mmHg (lit.)

mp

44-45 °C (lit.)

density

0.961 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCc1ccc(O)cc1

InChI

1S/C14H22O/c1-2-3-4-5-6-7-8-13-9-11-14(15)12-10-13/h9-12,15H,2-8H2,1H3

InChI key

NTDQQZYCCIDJRK-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carme Valls-Cantenys et al.
Journal of separation science, 37(24), 3706-3713 (2014-10-10)
Simple, precise, and low-cost methods for the simultaneous determination of phenolic endocrine disrupting compounds such as bisphenol A, trichlorophenol, pentachlorophenol, 4-nonylphenol, and 4-octylphenol in water samples were developed. The Direct, in situ derivatization methods are based on polydimethylsiloxane rod extraction
R White et al.
Endocrinology, 135(1), 175-182 (1994-07-01)
We show that a number of alkylphenolic compounds, used in a variety of commercial products and found in river water, are estrogenic in fish, birds, and mammals. 4-Octylphenol (OP), 4-nonylphenol, 4-nonylphenoxycarboxylic acid, and 4-nonylphenoldiethoxylate were each capable of stimulating vitellogenin
Tao Lv et al.
Journal of separation science, 37(19), 2757-2763 (2014-07-22)
A novel hyphenated method based on ultrasound-assisted dispersive liquid-liquid microextraction coupled to precolumn derivatization has been established for the simultaneous determination of bisphenol A, 4-octylphenol, and 4-nonylphenol by high-performance liquid chromatography with fluorescence detection. Different parameters that influence microextraction and
G G Kuiper et al.
Endocrinology, 139(10), 4252-4263 (1998-09-29)
The rat, mouse and human estrogen receptor (ER) exists as two subtypes, ER alpha and ER beta, which differ in the C-terminal ligand-binding domain and in the N-terminal transactivation domain. In this study, we investigated the estrogenic activity of environmental
Haidong Zhou et al.
Environmental monitoring and assessment, 161(1-4), 107-121 (2009-02-03)
Occurrence and fate of eight kinds of selected endocrine-disrupting compounds (EDCs) in three sewage treatment plants (STPs) of Beijing, China was investigated. These EDCs, composed of 4-octylphenol (4-OP), 4-n-nonylphenol (4-n-NP), bisphenol A (BPA), estrone (E1), 17alpha-estradiol (17alpha-E2), 17beta-estradiol (E2), estriol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service