Skip to Content
Merck
All Photos(2)

Documents

42088

Supelco

Bisphenol A

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2,2-Bis(4-hydroxyphenyl)propane, 4,4′-Isopropylidenediphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(C6H4OH)2
CAS Number:
Molecular Weight:
228.29
Beilstein:
1107700
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

220 °C/4 mmHg (lit.)

mp

158-159 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

InChI

1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3

InChI key

IISBACLAFKSPIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Ion Mobility: TWCCSN2 value of 158.8 Å2 [M-H]-

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 42088 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

440.6 °F - closed cup

Flash Point(C)

227 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Elin Engdahl et al.
International journal of molecular sciences, 22(11) (2021-06-03)
The breast cancer resistance protein (BCRP) is an important efflux transporter in the blood-brain barrier (BBB), protecting the brain from a wide range of substances. In this study, we investigated if BCRP function is affected by bisphenol A (BPA), a
Monika Rönn et al.
Chemosphere, 112, 42-48 (2014-07-23)
Since bisphenol A (BPA) has been shown to induce obesity in experimental studies, we explored the associations between BPA and fat mass, fat distribution and circulating levels of adiponectin, leptin and ghrelin in humans. In the Prospective Investigation of the
Tinne Geens et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(10), 3725-3740 (2012-08-15)
Due to the large number of applications of bisphenol-A (BPA), the human exposure routes are multiple. We aimed to review shortly the food and non-food sources of BPA, and to evaluate their contribution to the human exposure. Food sources discussed
Marie Tohmé et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(7), 3124-3133 (2014-04-20)
Bisphenol A (BPA) is an endocrine disruptor that displays estrogenic activity. Several reports suggest that BPA may have estrogen receptor-independent effects. In zebrafish, 50 μM BPA exposure induces otic vesicle abnormalities, including otolith aggregation. The purpose of this study was
R E FitzGerald et al.
Toxicology letters, 230(2), 368-374 (2014-05-17)
Bisphenol A (BPA) is one of the most widely used and extensively studied chemicals. Numerous studies have reported in vitro effects or animal adverse findings at BPA doses lower than the no observed adverse effect levels (NOAELs) established in regulatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service