Skip to Content
Merck
All Photos(1)

Key Documents

31732

Supelco

Abamectin

PESTANAL®, analytical standard

Synonym(s):

Avermectin B1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

CCC(C)C1O[C@@]2(C[C@@H]3CC(C\C=C(C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C6/COC7[C@H](O)C(C)=CC(C(=O)O3)[C@]67O)O2)C=C[C@@H]1C

InChI

1S/C48H72O14/c1-11-25(2)43-28(5)17-18-47(62-43)23-34-20-33(61-47)16-15-27(4)42(26(3)13-12-14-32-24-55-45-40(49)29(6)19-35(46(51)58-34)48(32,45)52)59-39-22-37(54-10)44(31(8)57-39)60-38-21-36(53-9)41(50)30(7)56-38/h12-15,17-19,25-26,28,30-31,33-45,49-50,52H,11,16,20-24H2,1-10H3/b13-12+,27-15+,32-14+/t25-,26-,28-,30-,31-,33+,34-,35?,36-,37-,38-,39-,40+,41-,42-,43+,44-,45+,47+,48+/m0/s1

InChI key

RRZXIRBKKLTSOM-RDKIRRGNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Abamectin (MK-0936) belongs to avermectin family. This family have a macrocycle as backbone with a spiroketal unit, a hexahydrobenzofuran unit and disaccharide substitute in general. MK-0936 is a natural fermenting product which can be produced by Streptomyces avermitilis.

Application

Abamectin has been used as standard in liquid chromatography–electrospray tandem mass spectrometry LC–ESI-MS–MS method for determination of abamectin residues in orange samples.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of abamectin and azadirachtin residues in orange samples by liquid chromatography?electrospray tandem mass spectrometry.
Pozo, O. J., et al.
Journal of Chromatography A, 992.1, 133-140 (2003)
Xiaojie Liu et al.
Frontiers in plant science, 8, 903-903 (2017-06-15)
The high structural diversity of plant metabolites suggests that interactions among them should be common. We investigated the effects of single metabolites and combinations of plant metabolites on insect herbivores. In particular we studied the interacting effects of pyrrolizidine alkaloid
Maria Edna Tenório Nunes et al.
Ecotoxicology (London, England), 21(4), 1063-1071 (2012-02-03)
Obtaining ecotoxicological data on pesticides in tropical regions is imperative for performing more realistic risk analysis, and avoidance tests have been proposed as a useful, fast and cost-effective tool. Therefore, the present study aimed to evaluate the avoidance behavior of
Ciler Celik-Ozenci et al.
Regulatory toxicology and pharmacology : RTP, 61(3), 310-317 (2011-09-29)
Despite the known adverse effects of abamectin pesticide, little is known about its action on male fertility. To explore the effects of exposure to abamectin on male fertility and its mechanism, low (1mg/kg/day) and high dose (4 mg/kg/day) abamectin were
Thomas Geurden et al.
Veterinary parasitology, 189(2-4), 308-316 (2012-05-09)
The objective of the present studies was to evaluate the efficacy of a combined formulation (Startect(®) Dual Active Oral Solution for Sheep, Pfizer Animal Health) of derquantel (DQL) and abamectin (ABA) for the treatment of: (1) sheep experimentally infected with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service