Skip to Content
Merck
All Photos(2)

Key Documents

32507

Supelco

Methoxyfenozide

PESTANAL®, analytical standard

Synonym(s):

3-Methoxy-2-methylbenzoic acid 2-(3,5-dimethylbenzoyl)-2-tert-butylhydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N2O3
CAS Number:
Molecular Weight:
368.47
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1cccc(C(=O)NN(C(=O)c2cc(C)cc(C)c2)C(C)(C)C)c1C

InChI

1S/C22H28N2O3/c1-14-11-15(2)13-17(12-14)21(26)24(22(4,5)6)23-20(25)18-9-8-10-19(27-7)16(18)3/h8-13H,1-7H3,(H,23,25)

InChI key

QCAWEPFNJXQPAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methoxyfenozide is a diacylhydrazine insecticide.

Application

Methoxyfenozide may be used as a reference standard for the determination of methoxyfenozide in vegetables using liquid chromatography-electrospray tandem mass spectrometry (LC-ESI-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hezhi Sun et al.
Journal of the science of food and agriculture, 100(6), 2358-2363 (2019-12-06)
Methoxyfenozide possesses efficacy against a variety of lepidopteron pests, including the major pests in cauliflower and tea, so it is of great importance to generalize the practical use of methoxyfenozide in the field. An efficient method was developed and validated
Lauren M Childs et al.
PLoS pathogens, 12(12), e1006060-e1006060 (2016-12-16)
The control of mosquito populations with insecticide treated bed nets and indoor residual sprays remains the cornerstone of malaria reduction and elimination programs. In light of widespread insecticide resistance in mosquitoes, however, alternative strategies for reducing transmission by the mosquito
William G Meikle et al.
PloS one, 14(3), e0204635-e0204635 (2019-03-29)
Methoxyfenozide is an insect growth regulator (IGR) commonly used in agriculture to simultaneously control pests and preserve beneficial insect populations; however, its impact on honey bees in not fully understood. We conducted field and laboratory experiments to investigate bee health
Faye Brown et al.
Insect biochemistry and molecular biology, 121, 103372-103372 (2020-04-11)
Insecticide based vector control tools such as insecticide treated bednets and indoor residual spraying represent the cornerstones of malaria control programs. Resistance to chemistries used in these programs is now widespread and represents a significant threat to the gains seen
Determination of tebufenozide and methoxyfenozide residues in vegetables using liquid chromatography-electrospray tandem mass spectrometry.
Lin D , et al.
Journal of Analytical Chemistry, 36(1), 87-90 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service