Skip to Content
Merck
All Photos(2)

Documents

74691

Supelco

Octadecane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CH3
CAS Number:
Molecular Weight:
254.49
Beilstein:
1740138
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

8.8 (vs air)

vapor pressure

1 mmHg ( 119 °C)

Assay

≥98.5% (GC)

autoignition temp.

455 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

317 °C (lit.)

mp

26-29 °C (lit.)
28-30 °C

density

0.777 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCCCCC

InChI

1S/C18H38/c1-3-5-7-9-11-13-15-17-18-16-14-12-10-8-6-4-2/h3-18H2,1-2H3

InChI key

RZJRJXONCZWCBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Octadecane has been used as a standard in the profiling of plant metabolites of Arabidopsis thaliana, via gas chromatography coupled with mass spectrometry method of analysis(GC-MS).
Octadecane may be used as an analytical reference standard for the determination of the analyte from hot-pressing of mixed-hardwood particleboard using chromatography techniques. It may be used as an internal standard to study the analysis of fatty acid composition of plasma glycerophospholipids using gas chromatography with flame ionization detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

330.8 °F - closed cup

Flash Point(C)

166 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolite Profiling of Mature Arabidopsis thaliana Seeds Using Gas Chromatography-Mass Spectrometry (GC-MS)
Cohen H, et al.
Bio-protocol, 6(21) (2016)
High-throughput analysis of fatty acid composition of plasma glycerophospholipids
Glaser C, et al.
Journal of Lipid Research, 51(1), 216-221 (2010)
Volatile organic emissions arising from the hot-pressing of mixed-hardwood particleboard
Jiang T, et al.
Forest products journal, 52(11), 66-77 (2002)
Abdul R Bhat et al.
Rapid communications in mass spectrometry : RCM, 24(24), 3547-3552 (2010-11-17)
We introduce a novel preconcentrating technique by using surface modification of palladium nanoparticles (Pd-NPs) with octadecane thiol (ODT) prepared in toluene for selective and sensitive extraction of proteins (insulin, ubiquitin, lysozyme) from a variety of real-world samples including pancreas, mushroom
Nadine Strehmel et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 871(2), 182-190 (2008-05-27)
The generation of retention index (RI) libraries is an expensive and time-consuming effort. Procedures for the transfer of RI properties between chromatography variants are, therefore, highly relevant for a shared use. The precision of RI determination and accuracy of RI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service