Skip to Content
Merck
All Photos(1)

Key Documents

31564

Supelco

Acrylonitrile

analytical standard

Synonym(s):

Vinyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCN
CAS Number:
Molecular Weight:
53.06
Beilstein:
605310
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

1.83 (vs air)

vapor pressure

86 mmHg ( 20 °C)

Assay

≥98% (GC)

autoignition temp.

897 °F

quality

stabilized

shelf life

limited shelf life, expiry date on the label

expl. lim.

17 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.391 (lit.)

bp

77 °C (lit.)

mp

−83 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C=CC#N

InChI

1S/C3H3N/c1-2-3-4/h2H,1H2

InChI key

NLHHRLWOUZZQLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acrylonitrile (AN) is a volatile liquid mostly used in manufacturing plastics and polymers.

Application

AN may have been used as standard during headspace gas chromatography for the determination of AN monomer in plastic containers and carbonated beverages.
Acrylonitrile is used in the manufacture of acrylic fibers, resins (acrylonitrile butadiene-styrene, styrene-acrylonitrile and others) and nitrile rubbers (butadiene-acrylonitrile).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

An industrial carcinogen that is a multisite carcinogen in rats and possibly carcinogenic to humans.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of acrylonitrile monomer in plastic packaging and beverages by headspace gas chromatography.
Gawell, GB-M.
Analyst, 104.1235, 106-110 (1979)
Role of cytochrome P450 2E1 in the metabolism of acrylamide and acrylonitrile in mice.
Sumner SC
Chemical Research in Toxicology, 12(11), 1110-1116 (1999)
Philip Cole et al.
Regulatory toxicology and pharmacology : RTP, 52(3), 342-351 (2008-10-18)
Several retrospective cohort epidemiology studies evaluated a number of health outcomes in workers exposed to acrylonitrile (AN). The epidemiology studies included in this review have been published since 1970 and were identified through Ovid and MEDLINE retrieval services using search
Acrylonitrile and human cancer--an overview.
D Coggon et al.
Scandinavian journal of work, environment & health, 24 Suppl 2, 81-82 (1998-08-26)
J Whysner et al.
Regulatory toxicology and pharmacology : RTP, 27(3), 217-239 (1998-08-07)
Acrylonitrile (ACN) exposure is associated with tumors in rat brain, Zymbal gland, and mammary gland. Adducts affecting base pairing were formed in isolated DNA exposed in vitro to the ACN metabolite cyanoethylene oxide (CNEO). DNA from liver, which is not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service