Skip to Content
Merck
All Photos(1)

Key Documents

76671

Supelco

Propionitrile

analytical standard

Synonym(s):

Ethyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CN
CAS Number:
Molecular Weight:
55.08
Beilstein:
773680
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.366 (lit.)
n20/D 1.366

bp

97 °C (lit.)

mp

−93 °C (lit.)

density

0.772 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCC#N

InChI

1S/C3H5N/c1-2-3-4/h2H2,1H3

InChI key

FVSKHRXBFJPNKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Propionitrile may be used as a mobile phase for the separation and elution of triglycerides in cocoa butter samples using reversed-phase high-performance liquid chromatography with refractive index detector and temperature programming.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved HPLC of triglycerides by special tempering procedures
Frede E
Chromatographia, 21(1), 29-36 (1986)
Sung Hee Joo et al.
Environmental science & technology, 41(4), 1288-1296 (2007-06-28)
The decreasing availability of pristine water supplies is prompting drinking water utilities to exploit waters impacted by wastewater effluents and agricultural runoff. As these waters feature elevated organic nitrogen concentrations, the pathways responsible for transformation of organic nitrogen into toxic
Jonna Frasor et al.
Endocrinology, 144(7), 3159-3166 (2003-06-18)
Estrogen is of great importance in the regulation of uterine function. The aim of this study was to examine the individual physiological roles of each of the two receptors for estradiol, estrogen receptor (ER) alpha and ERbeta, and their potential
M Cantarella et al.
Journal of industrial microbiology & biotechnology, 33(3), 208-214 (2005-03-02)
The biohydration of acrylonitrile, propionitrile and benzonitrile catalysed by the NHase activity contained in resting cells of Microbacterium imperiale CBS 498-74 was operated at 5, 10 and 20 degrees C in laboratory-scale batch and membrane bioreactors. The bioreactions were conducted
D Somjen et al.
Journal of cellular biochemistry, 112(2), 625-632 (2011-01-27)
In cultured human osteoblasts estradiol-17β (E2) modulated DNA synthesis, the specific activity of creatine kinase BB (CK), 12 and 15 lipoxygenase (LO) mRNA expression and formation of 12- and 15-hydroxyeicosatetraenoic acid (HETE). We now investigate the response of human bone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service