Skip to Content
Merck
All Photos(2)

Key Documents

94114

Supelco

Chlorpyrifos

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Chlorpyriphos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11Cl3NO3PS
CAS Number:
Molecular Weight:
350.59
Beilstein:
1545756
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1nc(Cl)c(Cl)cc1Cl

InChI

1S/C9H11Cl3NO3PS/c1-3-14-17(18,15-4-2)16-9-7(11)5-6(10)8(12)13-9/h5H,3-4H2,1-2H3

InChI key

SBPBAQFWLVIOKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J W Albers et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 2(4), 301-324 (1999-12-22)
This report summarizes the deliberations of an eight-member panel of scientists convened by Dow AgroSciences in cooperation with the U.S. Environmental Protection Agency (EPA). The panel was charged with evaluating the scientific literature on the health effects potentially associated with
Pamela J Mink et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 15(4), 281-316 (2012-05-11)
Chlorpyrifos (CPF) is one of the most widely used organophosphate insecticides in the United States. By December 2000, nearly all residential uses were voluntarily canceled, so that today, CPF is only used to control insect pests on a variety of
J Asselman et al.
Environmental pollution (Barking, Essex : 1987), 188, 56-63 (2014-02-21)
In aquatic ecosystems, mixtures of chemical and natural stressors can occur which may significantly complicate risk assessment approaches. Here, we show that effects of binary combinations of four different insecticides and Microcystis aeruginosa, a toxic cyanobacteria, on Daphnia pulex exhibited
D J Clegg et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 2(3), 257-279 (1999-08-03)
A panel of toxicology and medical experts was convened on 7-9 April 1997 to consider the available scientific literature on chlorpyrifos, both published and unpublished, to determine the acute and chronic toxicology reference dose (RfD). In the course of reviewing
Hyeong-Moo Shin et al.
Environmental science & technology, 47(2), 859-867 (2012-12-19)
Indoor residence times of semivolatile organic compounds (SVOCs) are a major and mostly unavailable input for residential exposure assessment. We calculated residence times for a suite of SVOCs using a fugacity model applied to residential environments. Residence times depend on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service