Skip to Content
Merck
All Photos(1)

Key Documents

07563

Supelco

4,4′-Difluorobenzophenone

TraceCERT®, certified reference material, 19F-qNMR Standard, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(FC6H4)2CO
CAS Number:
Molecular Weight:
218.20
Beilstein:
516231
EC Number:
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

100
300

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

qNMR: suitable

mp

102-105 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical

format

neat

SMILES string

Fc1ccc(cc1)C(=O)c2ccc(F)cc2

InChI

1S/C13H8F2O/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10/h1-8H

InChI key

LSQARZALBDFYQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out our entire range of quantitative NMR standards (qNMR standards)
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application


  • Enhancement of chromatographic analyses: This compound served as a standard in liquid chromatography coupled with photodiode array for the identification and quantification of co-eluting impurities in pharmaceuticals, showcasing its critical role in ensuring the purity and efficacy of medical products (Vecchietti et al., 2022).

  • Advanced pharmaceutical analysis: Employed in intelligent peak deconvolution in chromatography, 4,4′-Difluorobenzophenone is crucial for the detailed study of complex data matrices from pharmaceutical samples, aiding in the precise determination of compound structures and concentrations (Arase et al., 2016).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

305.6 °F - Pensky-Martens closed cup

Flash Point(C)

152 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service