Skip to Content
Merck
All Photos(2)

Key Documents

07038

Supelco

Dimethyl terephthalate

Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
Beilstein:
1107185
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

Standard for quantitative NMR
certified reference material
TraceCERT®

Quality Level

200
300

vapor density

1.04 (vs air)

vapor pressure

1.15 mmHg ( 93 °C)

description

qNMR Standard for DMSO-d6 (9.2 ppm)

product line

TraceCERT®

autoignition temp.

1058 °F

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable
qNMR: suitable

mp

139-141 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
pharmaceutical

format

neat

SMILES string

COC(=O)c1ccc(cc1)C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-3-5-8(6-4-7)10(12)14-2/h3-6H,1-2H3

InChI key

WOZVHXUHUFLZGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out our entire range of quantitative NMR standards (qNMR standards)
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Caution

Store under Argon.

Other Notes

Chemcial Shift: 8.1 and 3.9 ppm (chemical shifts may slightly vary depending on the experimental conditions)
Suitable NMR solvents: Acetonitrile, Chloroform, MeOD, DMSO-d6
Ion Mobility: TWCCSN2 value of 217.3 Å2 [M+Na]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 88173 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Recommended products

Check out ChemisTwin, our brand new online portal for identity confirmation of NMR spectra. A feature for quantitative NMR is in development. Learn more or reach out to us for a free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

309.2 °F - (External MSDS)

Flash Point(C)

154 °C - (External MSDS)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Claudio A Téllez et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(8-9), 2171-2180 (2004-07-14)
Fourier transform infrared and Raman spectra of dimethylterephthalate (DMT), as microcrystalline powder, have been investigated. The vibrational spectra were calculated using the AM1 and PM3 semi empirical procedures, and the Møller-Plesset (MP2/DZV), and the Becke-Lee, Yang and Parr gradient-corrected correlation
R I Goncharova et al.
TSitologiia i genetika, 36(1), 14-25 (2002-05-16)
In the mouse transplacental test, EMS induced micronuclei and disturbed haemopoiesis in female bone marrow and foetal liver. Dimethyl terephthalate at the tested dose was ineffective in pregnant females increasing however the level of these events in foetuses. Hence, both
Zhiqiang Guo et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(2), 337-340 (2007-02-28)
Two 10-hydroxybenzo[h]quinoline metal complexes, bis(10-hydroxybenzo[h]quinolinato) beryllium (Bebq2) and bis(10-hydroxybenzo[h]quinolinato)zincum (Znbq2), have been synthesized. The structure are characterized by 1HNMR, IR and so on. The photophysical processes of Bebq2 and Znbq2 have been carefully investigated by fluorescence spectra. The results show
S H Ganji et al.
Biodegradation, 6(1), 61-66 (1995-01-01)
Aspergillus niger (AG-1) metabolized dimethylterephthalate through monomethylterephthalate, terephthalate and protocatechuate. Degradation of dimethylterephthalate was followed by extraction of residual dimethylterephthalate from the spent medium. The quantitative UV analysis showed that 58% of the dimethylterephthalate supplement was taken up in 144
Minoru Genta et al.
Waste management (New York, N.Y.), 27(9), 1167-1177 (2006-08-18)
To apply PET depolymerization in supercritical methanol to commercial recycling, the benefits of supercritical methanol usage in PET depolymerization was investigated from the viewpoint of the reaction rate and energy demands. PET was depolymerized in a batch reactor at 573

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service