Skip to Content
Merck
All Photos(2)

Documents

41867

Supelco

Dimethyl sulfone

Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Methyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO2
CAS Number:
Molecular Weight:
94.13
Beilstein:
1737717
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

Standard for quantitative NMR
certified reference material
TraceCERT®

Quality Level

description

qNMR Standard for D2O and organic solvents (3.2 ppm)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
qNMR: suitable

bp

238 °C (lit.)

mp

107-109 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
pharmaceutical

format

neat

SMILES string

CS(C)(=O)=O

InChI

1S/C2H6O2S/c1-5(2,3)4/h1-2H3

InChI key

HHVIBTZHLRERCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfone is a solvent, which can dissolve a wide range of solutes and can be generally used for high temperature reactions.
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out our entire range of quantitative NMR standards (qNMR standards)

Application

Dimethyl sulfone has been used as internal standard, for the nuclear magnetic resonance spectrum studies of cocrystals containing API carbamazepine, synthesized using mechanochemical method of synthesis.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Chemical Shift: 3.2 ppm (chemical shifts may slightly vary depending on the experimental conditions)
suitable NMR solvents: D2O, DMSO-d6, CDCI3

Recommended products

Check out ChemisTwin, our brand new online portal for identity confirmation of NMR spectra. A feature for quantitative NMR is in development. Learn more or reach out to us for a free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Make and break-Facile synthesis of cocrystals and comprehensive dissolution studies
Batzdorf.L, et al.
Journal of Molecular Structure, 1133, 18-23 (2017)
Why are dimethyl sulfoxide and dimethyl sulfone such good solvents?
Clark T, et al.
Journal of Molecular Modeling, 14, 689-697 (2008)
Gaoqing Yuan et al.
Chemical communications (Cambridge, England), 48(60), 7513-7515 (2012-06-26)
With atmospheric oxygen as the oxidant, a novel copper(I)-catalyzed synthesis of aryl methyl sulfones from aryl halides and widely available DMSO is described. The procedure tolerates aryl halides with various functional groups (such as methoxy, acetyl, chloro, fluoro and nitro
M S Abdul Rasheed et al.
Poultry science, 99(12), 6559-6568 (2020-11-30)
A 28-day experiment was conducted in broilers to study the effects of feeding methylsulfonylmethane (MSM) and IL-10-neutralizing antibody from dried egg product (DEP) on the growth performance, immune responsivity, oxidative stress parameters, and gut health outcomes during a mild infection
S E Rose et al.
Magnetic resonance imaging, 18(1), 95-98 (2000-01-21)
We wish to report the detection of dimethyl sulfone (methylsulfonylmethane, C2H6O2S) in the brain of a normal 62-year-old male using in vivo proton magnetic resonance spectroscopy. The presence of this exogenous metabolite resulted from ingestion of a dietary supplement containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service