53396
2,4-Dichlorobenzotrifluoride
TraceCERT®, certified reference material, 19F-qNMR Standard, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2,4-Dichloro-α,α,α-trifluorotoluene
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
100
300
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
technique(s)
qNMR: suitable
refractive index
n20/D 1.481 (lit.)
bp
117-118 °C (lit.)
density
1.484 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
FC(F)(F)c1ccc(Cl)cc1Cl
InChI
1S/C7H3Cl2F3/c8-4-1-2-5(6(9)3-4)7(10,11)12/h1-3H
InChI key
KALSHRGEFLVFHE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Check out our entire range of quantitative NMR standards (qNMR standards)
Application
- Environmental remediation and analysis: The study conducted by Chen et al. (2017) investigates the degradation of 3,4-dichlorobenzotrifluoride using zirconia-coated magnetite magnetic nanoparticles in a Fenton-like process, suggesting its relevance in environmental remediation technologies for halogenated aromatic compounds (Chen et al., 2017).
- Groundwater contamination monitoring: Research by Lava et al. (2013) focuses on the analytical determination of benzotrifluoride derivatives, including 2,4-dichlorobenzotrifluoride, in groundwater. This study underlines the compound′s significance as an analytical standard in monitoring environmental contamination and assessing the impact of industrial pollutants (Lava et al., 2013).
Recommended products
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4 - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
161.6 °F - closed cup
Flash Point(C)
72 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service