Skip to Content
Merck
All Photos(1)

Key Documents

04502

Sigma-Aldrich

Zinc phosphide

≥19% active phoshor (P) basis, powder

Synonym(s):

Trizinc diphosphide, Zn3P2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn3P2
CAS Number:
Molecular Weight:
258.12
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

powder

composition

active phoshor (P), ≥19%

reaction suitability

core: zinc
reagent type: catalyst

storage temp.

room temp

SMILES string

[Zn]=P[Zn]P=[Zn]

InChI

1S/2P.3Zn

InChI key

NQDYSWQRWWTVJU-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han Sung Kim et al.
Nano letters, 10(5), 1682-1691 (2010-04-15)
Electron tomography and high-resolution transmission electron microscopy were used to characterize the unique three-dimensional (3D) structures of twinned Zn(3)P(2) (tetragonal) and InAs (zinc blende) nanowires synthesized by the vapor transport method. The Zn(3)P(2) nanowires adopt a unique superlattice structure that
Gary W Witmer et al.
Bulletin of environmental contamination and toxicology, 84(6), 698-702 (2010-05-01)
Nutria, Myocastor coypus, populations must be reduced when they cause substantial wetland damage. Control can include the rodenticide zinc phosphide, but the potential impacts to American alligators, Alligator mississippiensis, must be assessed. The mean amount of zinc phosphide per nutria
Pavol Bakoss et al.
Annals of agricultural and environmental medicine : AAEM, 14(1), 51-56 (2007-07-28)
The purpose of the study was to suppress a known natural focus of field fever exerting an influence on animal hosts of leptospires--small terrestrial mammals (s.t.m.) by rodenticide. After repeated application of the zinc phosphide rodenticide, the s.t.m. were regularly
Comment on Aluminum and zinc phosphide poisoning.
Omid Mehrpour et al.
Clinical toxicology (Philadelphia, Pa.), 47(8), 838-839 (2009-09-26)
Jens Jacob et al.
Pest management science, 66(2), 132-136 (2009-10-03)
Minimising the concentration of active ingredient in rodenticide bait is desirable economically and for the protection of the agroecosystem. This study aimed to identify a zinc phosphide concentration that balances palatability and efficacy for common vole (Microtus arvalis Pall.) management

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service