Skip to Content
Merck
All Photos(4)

Key Documents

A4915

Sigma-Aldrich

Ammonium sulfate

ACS reagent, ≥99.0%

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38030408
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0%
grade:
ACS reagent
form:
powder or crystals
solubility:
water: soluble 132 g/L at 20 °C

grade

ACS reagent

Quality Level

Assay

≥99.0%

form

powder or crystals

impurities

≤0.005% Insoluble matter

ign. residue

≤0.005%

pH

5.0-6.0 (25 °C, 5%)

mp

>280 °C (dec.) (lit.)

solubility

water: soluble 132 g/L at 20 °C

density

1.77 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm
nitrate (NO3-): ≤0.001%
phosphate (PO43-): ≤5 ppm

cation traces

Fe: ≤5 ppm
heavy metals: ≤5 ppm (by ICP-OES)

storage temp.

room temp

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ammonium sulfate is an ammonium salt commonly used as fertilizer. It can be synthesized by neutralizing sulfuric acid with ammonia. It can enhance the phytotoxicity of several herbicides like glyphosates. Ammonium sulfate is formed as a co-product during the manufacture of caprolactam, a precursor of nylon.

Application

Ammonium sulfate was used in methyl cellulose precipitable tannin assay to evaluate wine tannin in finished wine samples. It was also used in making externally mixed aerosols for analyzing the 1 × 3 tandem differential mobility analyzer (1 × 3-TDMA), which is mainly employed for determining the irreversibility of the hygroscopic growth factor of aerosol particles.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Substitutes for Ammonium Sulfate as Additives with Glyphosate and Glufosinate.
Pratt D, et al.
Weed Technology, 17(3), 576-581 (2003)
The 1-by-3 tandem differential mobility analyzer for measurement of the irreversibility of the hygroscopic growth factor.
Rosenoern T, et al.
Aerosol Science and Technology, 43(7), 641-652 (2009)
Ammonium Compounds.
Weston CW, et al.
Kirk-Othmer Encyclopedia of Chemical Technology (2003)
Industrial catalytic aspects of the synthesis of monomers for nylon production.
Bellussi G and Perego C.
CATTECH, 4(1), 4-16 (2000)
Influence of yeast strain on Shiraz wine quality indicators.
Holt H, et al.
International Journal of Food Microbiology, 165(3), 302-311 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service