Skip to Content
Merck
All Photos(2)

Key Documents

343242

Sigma-Aldrich

Phosphorus, red

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
P
CAS Number:
Molecular Weight:
30.97
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

vapor density

0.02 (vs air)

Quality Level

vapor pressure

0.03 mmHg ( 21 °C)
1 mmHg ( 76.6 °C)

Assay

≥99.99% trace metals basis

form

powder or chunks

autoignition temp.

500 °F

resistivity

10 μΩ-cm, 20°C

mp

416 °C (subl.) (lit.)

density

2.34 g/mL at 25 °C (lit.)

SMILES string

[P]

InChI

1S/P

InChI key

OAICVXFJPJFONN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Recycled Graphite from Spent Lithium-Ion Batteries as a Conductive Framework Directly Applied in Red Phosphorus-Based Anodes.: This study explores using recycled graphite from spent lithium-ion batteries to create a conductive framework for red phosphorus-based anodes, enhancing their performance and sustainability in battery applications (Huang et al., 2023).

  • P-Doping a Porous Carbon Host Promotes the Lithium Storage Performance of Red Phosphorus.: This research demonstrates that P-doping of a porous carbon host significantly improves the lithium storage performance of red phosphorus, offering a promising approach for high-capacity battery anodes (Han et al., 2023).

  • Laser-Ablated Red Phosphorus on Carbon Nanotube Film for Accelerating Polysulfide Conversion toward High-Performance and Flexible Lithium-Sulfur Batteries.: The study introduces a laser-ablation technique to incorporate red phosphorus onto carbon nanotube films, enhancing polysulfide conversion and overall battery performance (Lee et al., 2021).

  • Green, Template-Less Synthesis of Honeycomb-like Porous Micron-Sized Red Phosphorus for High-Performance Lithium Storage.: This paper presents a green, template-less synthesis method for creating honeycomb-like porous red phosphorus, leading to improved lithium storage capabilities for battery applications (Zhu et al., 2021).


Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Aloysious Aravinthan et al.
Experimental gerontology, 60, 37-45 (2014-09-23)
Hepatocyte senescence is associated closely with fibrosis stage and an adverse outcome in chronic liver disease, but it is uncertain whether there is a causal relation with clinical manifestations of chronic liver disease, which was the subject of this study
Jacques C Finlay et al.
Science (New York, N.Y.), 342(6155), 247-250 (2013-10-12)
Human activities have increased the availability of reactive nitrogen in many ecosystems, leading to negative impacts on human health, biodiversity, and water quality. Freshwater ecosystems, including lakes, streams, and wetlands, are a large global sink for reactive nitrogen, but factors
Yuki Fujita et al.
Nature, 505(7481), 82-86 (2013-11-19)
Plant species diversity in Eurasian wetlands and grasslands depends not only on productivity but also on the relative availability of nutrients, particularly of nitrogen and phosphorus. Here we show that the impacts of nitrogen:phosphorus stoichiometry on plant species richness can
Manuel Delgado-Baquerizo et al.
Nature, 502(7473), 672-676 (2013-11-01)
The biogeochemical cycles of carbon (C), nitrogen (N) and phosphorus (P) are interlinked by primary production, respiration and decomposition in terrestrial ecosystems. It has been suggested that the C, N and P cycles could become uncoupled under rapid climate change
Bon-Chul Koo et al.
Science (New York, N.Y.), 342(6164), 1346-1348 (2013-12-18)
Phosphorus ((31)P), which is essential for life, is thought to be synthesized in massive stars and dispersed into interstellar space when these stars explode as supernovae (SNe). Here, we report on near-infrared spectroscopic observations of the young SN remnant Cassiopeia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service