Skip to Content
Merck

Skip To

02474

4-Methyl-2-pentanone

analytical standard

Synonym(s):

Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2CHCH2COCH3
CAS Number:
Molecular Weight:
100.16
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
203-550-1
Beilstein/REAXYS Number:
605399
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

vapor density

3.5 (vs air)

assay

≥99.7% (GC)

autoignition temp.

840 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

1.2-8 %, 93 °F

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.395 (lit.), n20/D 1.396

bp

117-118 °C

mp

−80 °C (lit.)

density

0.801 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)CC(C)=O

InChI

1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3

InChI key

NTIZESTWPVYFNL-UHFFFAOYSA-N

General description

4-Methyl-2-pentanone (Methyl isobutyl ketone (MBK)) is an organic solvent widely used in various industrial applications, and exists as a clear colorless liquid with a pleasant odor. It is occasionally found as a volatile component of urine making it a biological marker of occupational exposure to this solvent. It is used as is used as a solvent for gums, resins, paints, varnishes, lacquers, and nitrocellulose.

Application

MBK may have been used as internal standard in GC/MS analysis for determining off-flavors in milk extracted using solid-phase microextraction, mass spectrometry, and multivariate analysis (SPME-MS-MVA). 4-Methyl-2-pentanone may be used as an analytical reference standard for the quantification of the analyte in hydro-alcoholic matrices using high-performance liquid chromatography with diode array detection.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W2731045377131.06146
application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

application(s)

flavors and fragrances

application(s)

-

application(s)

-

grade

analytical standard

grade

Kosher

grade

-

grade

ACS reagent

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

format

neat

format

-

format

-

format

-

assay

≥99.7% (GC)

assay

≥99%

assay

≥99%

assay

≥99% (GC)

refractive index

n20/D 1.395 (lit.), n20/D 1.396

refractive index

n20/D 1.395 (lit.)

refractive index

n20/D 1.395 (lit.)

refractive index

n20/D 1.395 (lit.)


Still not finding the right product?

Explore all of our products under 4-Methyl-2-pentanone


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

57.2 °F - closed cup

flash_point_c

14 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



SPME-MS-MVA as an electronic nose for the study of off-flavors in milk.
Marsili RT
Journal of Agricultural and Food Chemistry, 47(2), 648-654 (1999)
HPLC-DAD analysis of ketones as their 2, 4-dinitrophenylhydrazones in Brazilian sugar-cane spirits and rum
Cardoso D, et al.
J. Food Compos. Anal., 16(5), 563-573 (2003)
Ginette Truchon et al.
Journal of occupational and environmental hygiene, 3(3), 137-143 (2006-02-09)
Compartmental and physiologically based toxicokinetic modeling coupled with Monte Carlo simulation were used to quantify the impact of biological variability (physiological, biochemical, and anatomic parameters) on the values of a series of bio-indicators of metal and organic industrial chemical exposures.



Global Trade Item Number

SKUGTIN
213047-50G04061838772572
02474-5ML04061838617255
EHNC-024741-50UG04065265332675
EHNC-024741-20UG04065265332668
EHNC024741-20UG04061841396888
EHNC024741-50UG04061841396895

Questions

Reviews

No rating value

Active Filters