Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-Methyl-2-pentanone
Synonym(s):
Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone
Linear Formula:
(CH
3
)
2
CHCH
2
COCH
3
CAS Number:
108-10-1
Molecular Weight:
100.16
EC Number:
203-550-1
Products
Product Category
solvents (3)
aroma chemicals (1)
Brand
Sigma-Aldrich (4)
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
ACS reagent (1)
Kosher (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
293261
4-Methyl-2-pentanone,
suitable for HPLC, ≥99.5%
Expand
View Pricing
360511
4-Methyl-2-pentanone,
ACS reagent, ≥98.5%
Expand
View Pricing
537713
4-Methyl-2-pentanone,
≥99%
Expand
View Pricing
W273104
4-Methyl-2-pentanone,
≥99%, FCC
Expand
View Pricing
02474
4-Methyl-2-pentanone,
analytical standard
Expand
View Pricing