Skip to Content
Merck
All Photos(1)

Documents

00738

Sigma-Aldrich

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate

for electrochemistry, ≥98.0% (T)

Synonym(s):

EMIM Otf

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11F3N2O3S
CAS Number:
Molecular Weight:
260.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

for electrochemistry

Quality Level

Assay

≥98.0% (T)

form

liquid

impurities

≤0.1% water

refractive index

n20/D 1.435 (lit.)

bp

>350 °C (lit.)

density

1.387 g/mL at 25 °C (lit.)

anion traces

bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤10 mg/kg
fluoride (F-): ≤100 mg/kg

SMILES string

CCn1cc[n+](C)c1.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/C6H11N2.CHF3O3S/c1-3-8-5-4-7(2)6-8;2-1(3,4)8(5,6)7/h4-6H,3H2,1-2H3;(H,5,6,7)/q+1;/p-1

InChI key

ZPTRYWVRCNOTAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

425.3 °F - closed cup

Flash Point(C)

218.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service