Skip to Content
Merck
All Photos(1)

Documents

89320

Sigma-Aldrich

1-Hexyl-3-methylimidazolium hexafluorophosphate

≥97.0% (HPLC)

Synonym(s):

HMIMPF6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19F6N2P
CAS Number:
Molecular Weight:
312.24
Beilstein:
8955409
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

electrochemical window 5.5 V

Quality Level

Assay

≥97.0% (HPLC)

form

liquid

refractive index

n20/D 1.419

SMILES string

F[P-](F)(F)(F)(F)F.CCCCCCn1cc[n+](C)c1

InChI

1S/C10H19N2.F6P/c1-3-4-5-6-7-12-9-8-11(2)10-12;1-7(2,3,4,5)6/h8-10H,3-7H2,1-2H3;/q+1;-1

InChI key

YPWSSSRXUOQNMQ-UHFFFAOYSA-N

General description

1-Hexyl-3-methylimidazolium hexafluorophosphate is a water-immiscible imidazolium-type room temperature ionic liquid (RTIL) suitable for electrochemistry.
https://www.sigmaaldrich.com/content/dam/sigma-aldrich/materials-science/material-matters/material_matters_v4n4.pdf#page=18

Application

HMIMPF6 can be used as a chelate extraction solvent for divalent metal cations using 4,4,4-trifluoro-1-(2-thienyl)-1,3-butanedione (thenoyltrifluoroacetone, Htta) as an extractant.

Other Notes

Ionic liquid employed in Friedel-Crafts alkylation of aromatic compounds; polarity of salt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C.E. Song et al.
Chemical Communications (Cambridge, England), 1695-1695 (2000)
Arezou Taghvimi et al.
Journal of separation science, 42(2), 582-590 (2018-11-11)
Ionic liquid carbon-coated magnetic nanoparticles were successfully applied as an adsorbent in a mixed hemimicelle magnetic dispersive solid-phase extraction method for the determination of tramadol from urine samples coupled with high-performance liquid chromatography with UV-vis detection. The significant parameters affect
Use of 1-alkyl-3-methylimidazolium hexafluorophosphate room temperature ionic liquids as chelate extraction solvent with 4, 4, 4-trifluoro-1-(2-thienyl)-1, 3-butanedione.
Hirayama N, et al.
Talanta, 65(1), 255-260 (2005)
Hassan Karimi-Maleh et al.
Journal of colloid and interface science, 554, 603-610 (2019-07-23)
A voltammetric sensor was fabricated for the determination of 2-phenylphenol (2-PP) as a water pollutant. The sensor was prepared by modifying carbon paste electrode (CPE) with a composite of Fe3O4 nanoparticles (Fe3O4-NPs) and ionic liquid (n-hexyl-3-methylimidazolium hexafluorophosphate; (HMPF6)). The Fe3O4-NPs
Viscosity of 1-hexyl-3-methylimidazolium hexafluorophosphate and 1-octyl-3-methylimidazolium hexafluorophosphate at high pressure
Tomida, Daisuke, et al.
Journal of Chemical and Engineering Data, 52.2, 577-579 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service