Skip to Content
Merck
All Photos(1)

Key Documents

711748

Sigma-Aldrich

1-Butyl-3-methylimidazolium tetrafluoroborate

≥98%

Synonym(s):

BMIMBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15BF4N2
CAS Number:
Molecular Weight:
226.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

liquid

impurities

≤0.5% water

density

1.21 g/mL at 20 °C (lit.)

SMILES string

F[B-](F)(F)F.CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.BF4/c1-3-4-5-10-7-6-9(2)8-10;2-1(3,4)5/h6-8H,3-5H2,1-2H3;/q+1;-1

InChI key

LSBXQLQATZTAPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Butyl-3-methylimidazolium tetrafluoroborate is a hydrophilic room temperature ionic liquid (RTIL).

Application

1-Butyl-3-methylimidazolium tetrafluoroborate ([BMIM]BF4) can be used:  
  • As a reaction medium for the preparation NH4TiOF3 mesocrystals, which are converted into TiO2 based nanostructures.        
  • As a working fluid along with 2,2,2-trifluoroethanol in absorption heat pumps or chillers.       
  • As an electrolyte in lithium-ion batteries and double layer capacitors.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

550.4 °F

Flash Point(C)

288 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nora Aranburu et al.
Polymers, 11(3) (2019-04-10)
Biobased thermoplastic polyurethane (bTPU)/unmodified graphene (GR) nanocomposites (NCs) were obtained by melt-mixing in a lab-scaled conventional twin-screw extruder. Alternatively, GR was also modified with an ionic liquid (GR-IL) using a simple preparation method with the aim of improving the dispersion
Xiaojing Lv et al.
Nanotechnology, 31(35), 355201-355201 (2020-05-15)
TiO2 nanowires with high transparency and good ion storage capacity were explored as the charge-balancing layers for assembling electrochromic devices (ECDs). Increase thickness of TiO2 nanowires layer lowers the driving potential of the entire ECDs accompanied with reduced potential at
Effects of hydrophilic room-temperature ionic liquid 1-butyl-3-methylimidazolium tetrafluoroborate on direct electrochemistry and bioelectrocatalysis of heme proteins entrapped in agarose hydrogel films.
Wang SF, et al.
Electrochemical Communications, 9(7), 1709-1714 (2007)
Is the mixture of 1-ethyl-3-methylimidazolium tetrafluoroborate and 1-butyl-3-methylimidazolium tetrafluoroborate applicable as electrolyte in electrical double layer capacitors?
Palm R, et al.
Electrochemical Communications, 22, 203-206 (2012)
Preparation and properties of nanostructure anatase TiO2 monoliths using 1-butyl-3-methylimidazolium tetrafluoroborate room-temperature ionic liquids as template solvents.
Liu Y, et al.
Crystal Growth & Design, 5(4), 1643-1649 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service