Skip to Content
Merck
All Photos(7)

Documents

PFORCEKB

Roche

KAPA PROBE FORCE

2 ×, Universal

Synonym(s):

qPCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

usage

sufficient for 100 reactions
20 μL sufficient for 100 reactions
sufficient for 500 reactions
20 μL sufficient for 500 reactions

shelf life

≤12 mo.

feature

dNTPs included
hotstart

packaging

kit of 1 mL (100 x 20 μL rxn; KK4300)
kit of 5 mL (500 x 20 μL rxn; KK4301)

manufacturer/tradename

Roche

concentration

2 ×

technique(s)

qPCR: suitable

input

crude DNA

detection method

probe-based

storage temp.

−20°C

General description

KAPA PROBE FORCE is a highly inhibitor resistant qPCR master mix that removes the need for DNA purification, enabling streamlined sample-to-Cq workflows. The master mix contains a third generation DNA polymerase evolved to overcome blood, tissue, and plant PCR inhibitors. Crude samples can now be analyzed with comparable accuracy, reproducibility, and sensitivity as purified DNA using KAPA PROBE FORCE.
  • Direct qPCR from crude blood, tissue, and plant extracts
  • Sample-to-Cq workflows in <1 hour
  • High efficiency for accurate, reproducible, and sensitive results
  • Superior tolerance to carry-over inhibitors
  • Multiplex compatibility with crude extracts

Application

KAPA PROBE FORCE has been used for:
  • GMO testing
  • Mouse transgenics
  • SNP genotyping
  • Food/water pathogen detection
  • Infectious disease research
  • Cancer research
  • DNA quantification
  • Quantitative polymerase chain reaction (qPCR)
  • Digital droplet PCR
  • For amplification of templates directly from cDNA synthesis reactions

Features and Benefits

Streamline sample-to-Cq workflows in <1 hour:
  • Eliminate the time and cost of sample purification by amplifying directly from crude samples
  • Analyze a wide range of sample types including whole blood, cells, mouse tails, FFPE, leaf, stem, seed, and soil

Generate accurate and reproducible results:
  • Kits include a third-generation DNA polymerase, evolved for robust target amplification and detection
  • Enzyme maintains high reaction efficiency in the presence of PCR inhibitors for reliable data generation

Break through high levels of qPCR inhibitors:
  • Achieve greater levels of sensitivity for inhibited blood, tissue, and plant samples
  • Convert purified DNA assays to crude workflows without observable Cq delays

Multiplex crude samples efficiently:
  • Accelerate genotyping analysis with single reaction allelic discrimination of crude DNA extracts
  • Maximize data collection from precious samples, increase throughput, and reduce costs

Quick Notes:
  • This kit contains the KAPA3G HotStart DNA Polymerase enzyme, enabling probe-based qPCR for both routine and challenging sample types.
  • Initial denaturation of 3 min at 98°C is recommended to ensure complete denaturation of complex target DNA. A 5-min denaturation time may be required for some crude samples.
  • For two-step cycling, use a 20-sec combined annealing/extension/data acquisition at 60°C as a first approach.
  • A 10-sec annealing/extension/data acquisition time may be used with most assays, but this must be determined empirically.
  • For crude samples, the amount of sample in the reaction may be reduced to improve performance, but this must be determined empirically.

Quality

KAPA PROBE FORCE qPCR Master Mix Universal is subjected to stringent functional quality control, free of detectable contaminating exo- and endonuclease activities, and meets strict requirements with respect to DNA contamination.

Preparation Note

ROX reference dye is sensitive to light exposure. Always ensure that the product has been fully thawed and mixed before use. Avoid repeated freezing and thawing.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • KAPA3G HotStart® DNA Polymerase

  • dNTPs (including dUTP)

  • MgCl2 4.5 mM at 1X

  • ROX reference dye

  • stabilizers

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helga Bergholtz et al.
Cancer reports (Hoboken, N.J.), 3(3), e1248-e1248 (2020-07-17)
Ductal carcinoma in situ (DCIS) comprises a diverse group of preinvasive lesions in the breast and poses a considerable clinical challenge due to lack of markers of progression. Genomic alterations are to a large extent similar in DCIS and invasive
Development of genotyping method for functionally relevant variants of cytochromes P450 in cynomolgus macaques.
Uno Y, et al.
Journal of Veterinary Pharmacology and Therapeutics, 41(1), e30-e34 (2018)
Clarity? digital PCR system: a novel platform for absolute quantification of nucleic acids.
Low H, et al.
Analytical and Bioanalytical Chemistry, 409(7), 1869-1875 (2017)
L F S Leandro et al.
Plant disease, 102(9), 1748-1758 (2018-08-21)
Current management of sudden death syndrome (SDS) of soybean, caused by Fusarium virguliforme, focuses on planting resistant varieties and improving soil drainage; however, these measures are not completely effective. A 6-year study evaluated the effects of cropping system diversification on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service