Skip to Content
Merck
All Photos(5)

Documents

2GRHSRMKB

Roche

KAPA2G Robust HotStart ReadyMix

2 × ( contains 2 mM MgCl2 at 1×)

Synonym(s):

PCR, hotstart PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

usage

sufficient for 100 reactions
sufficient for 500 reactions

Quality Level

shelf life

≤12 mo.

feature

Difficult Templates/Specialty Enzymes PCR
dNTPs included
hotstart

packaging

kit of 1.25 mL (100 x 25 μL rxn; KK5701)
kit of 6.25 mL (500 x 25 μL rxn; KK5702)

manufacturer/tradename

Roche

concentration

2 × ( contains 2 mM MgCl2 at 1×)

technique(s)

PCR: suitable

input

crude DNA

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The second-generation KAPA2G Robust DNA Polymerase was evolved to solve inconsistent amplification across a broad range of amplicon types (GC- and AT-rich). KAPA2G Robust HotStart® ReadyMix enables higher processivity and specific activity, which translates to robust PCR performance, high sensitivity, and improved tolerance to common inhibitors. The high performance of the KAPA2G Robust DNA Polymerase is ideally suited for challenging PCR applications and difficult samples, eliminating the need for optimization using multiple enzymes and protocols.

Application

KAPA2G Robust HotStart® ReadyMix has been used for the:
  • amplification of GC- or AT-rich templates
  • aplification of templates containing common polymerase chain reaction (PCR) inhibitors e.g. salts, urea, SDS, or ethanol
  • crude sample PCR e.g. buccal swabs, cultured mammalian, yeast, or bacterial cells
  • colony PCR
  • DNA extraction and amplification
  • mutation screening
  • methylation-specific PCR (MSP) amplification
  • genotyping-PCR
  • PCR-based sequencing

Biochem/physiol Actions

DNA fragments generated with KAPA2G Robust HotStart® DNA Polymerase have the same characteristics as DNA fragments generated with wild-type Taq DNA polymerase and may be used for routine downstream analyses or applications, including restriction enzyme digestion, cloning, and sequencing. Like wild-type Taq, KAPA2G Robust HotStart has 5′→3′ polymerase and 5′→3′ exonuclease activities, but no 3′→5′ exonuclease (proofreading) activity. The fidelity of KAPA2G Robust HotStart is similar to that of wild-type Taq; it has an error rate of approximately 1 error per 1.7 x 105 nucleotides incorporated. PCR products generated by KAPA2G Robust HotStart are 3′-dA-tailed and may be cloned into TA cloning vectors.

Features and Benefits

Efficient amplification of GC- and AT-rich targets
  • Robust performance across a wide range of GC- and AT-rich templates
  • Increased PCR success rates

Improved tolerance to common PCR inhibitors
  • Efficient amplification from crude samples
  • Higher yield and sensitivity per unit of enzyme

Unrivalled performance in colony PCR
  • Higher yields and improved consistency of PCR direct from E. coli and yeast cells

Quick Notes:
  • KAPA2G Robust HotStart ReadyMix PCR Kits contain KAPA2G Robust DNA Polymerase, engineered for high processivity and inhibitor tolerance.
  • Both purified genomic DNA and crude samples (e.g. colony PCR) can be used as template.
  • Use 15 sec/kb extension time per cycle, and increase to 30-60 sec/kb for difficult amplicons or templates.
  • KAPA2G Robust HotStart ReadyMix contains 2 mM MgCl2 at 1X.
  • KAPA2G Robust HotStart ReadyMix with dye includes two inert tracking dyes to allow direct loading onto agarose gels.
  • KAPA2G Robust HotStart ReadyMix is ideal for single-protocol PCR, i.e. amplification of fragments varying from 25–65% GC, up to 1 kb in size, using a single reaction setup and cycling protocol.
  • Reaction products are 3′-dA-tailed and may be cloned into TA cloning vectors.

Quality

Each batch of KAPA2G Robust HotStart DNA Polymerase is confirmed to contain <2% contaminating protein (Agilent Protein 230 Assay). KAPA2G Robust HotStart ReadyMix PCR Kits are subjected to stringent quality control tests, are free of contaminating exo- and endonuclease activity, and meet strict requirements with respect to DNA contamination levels.

Preparation Note

Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for long-term storage. Provided that all components have been handled carefully and not contaminated, the kit is not expected to be compromised if left (unintentionally) at room temperature for a short period of time (up to 3 days). Long-term storage at room temperature and 4°C is not recommended. Please note that reagents stored at temperatures above -20°C are more prone to degradation when contaminated during use, and therefore storage at such temperatures is at the user′s own risk.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

HOTSTART is a registered trademark of Molecular BioProducts, Inc.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • KAPA2G Robust HotStart® DNA Polymerase 1U per 25 µL reaction

  • MgCl2 2mM at 1X

  • dNTP Mix 0.2 mM of each dNTP at 1X

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massively parallel characterization of CYP2C9 variant enzyme activity and abundance
Amorosi, et al.
American Journal of Human Genetics, 108, 1735-1751 (2021)
ssAAVs containing cassettes encoding SaCas9 and guides targeting hepatitis B virus inactivate replication of the virus in cultured cells
Scott, et al.
Scientific Reports, 7 (2017)
Disseminated Human Subarachnoid Coenurosis
Labuschagne, et al.
Tropical medicine and infectious disease, 7 (2022)
Supplementation with N-3 long-chain polyunsaturated fatty acids or olive oil in men and women with renal disease induces differential changes in the DNA methylation of FADS2 and ELOVL5 in peripheral blood mononuclear cells.
Hoile S P, et al.
PLoS ONE, 9(10), e109896-e109896 (2014)
Investigation of Oxtr-expressing Neurons Projecting to Nucleus Accumbens using Oxtr-ires-Cre Knock-in prairie Voles (Microtus ochrogaster)
Horie K, et al.
Neuroscience journal (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service