Skip to Content
Merck
All Photos(3)

Key Documents

EXPPCRKB

Roche

KAPA Express Extract

with PCR ReadyMix

Synonym(s):

PCR, PCR extraction

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

usage

sufficient for 100 reactions
sufficient for 500 reactions

shelf life

≤12 mo.

packaging

pkg of 1000 U (KK7152)
pkg of 200 U (KK7151)

manufacturer/tradename

Roche

technique(s)

DNA extraction: suitable
PCR: suitable

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

KAPA Express Extract with polymerase chain reaction (PCR) ReadyMix is a novel thermostable protease and buffer system that allows for the extraction of PCR-ready DNA in less than 15 minutes. DNA extractions are conveniently performed in a single-tube thereby greatly reducing the risk of sample loss and contamination.
The combination of KAPA Express Extract and KAPA2G Robust HotStart® ReadyMix provides a high performance solution for rapid DNA extraction and consistent downstream amplification.

Application

KAPA Express Extract with PCR ReadyMix is ideally suited for the extraction of PCR-ready DNA from, but not limited to, the following sample types:
  • Human tissue (formalin-fixed paraffin-embedded (FFPE) samples; blood collected in EDTA tubes or on collection cards; buccal swabs; hair follicles; forensic samples)
  • Animal samples (ear or tail clippings; hair follicles; blood; bone marrow; dried or fresh tissue from mouse and other mammals)
  • Fish tissues (fin punches; fresh tissue; cold and hot smoked canned samples; ethanol-preserved samples)
  • Insects (crushed)
  • Bird feathers (calamus fragments).

Features and Benefits

Key Features:
  • Rapid extraction protocol: PCR-ready DNA in 15 minutes
  • Versatility: single kit optimized for a variety of sample types
  • Single-tube reaction minimizes risk of contamination
  • DNA extracted with KAPA Express Extract is compatible with any PCR system

Rapid DNA extraction from a variety of sample types:
  • From sample to PCR in less than 15 minutes
  • Successful extraction from a variety of tissue types, including buccal swabs, hair follicles, fish tissue, and bird feathers

Increased PCR success from formalin-fixed paraffin-embedded (FFPE) samples or tissues:
  • Rapid DNA extraction and consistent downstream amplification of challenging sample types

Routine DNA extraction from blood sample types:
  • Efficient extraction from “Guthrie”cards, FTA Elute cards, and FTA cards

Quick Notes:
  • Extract PCR-ready DNA from different tissue types in a quick and simple, single-tube protocol (15–20 min), without the need for hazardous chemicals or multiple washing steps.
  • Significantly reduced turnaround times and risk of sample loss and contamination.
  • Use 1 μL DNA extract per 25 μL PCR. Depending on the sample type, the extract may have to be diluted prior to PCR.
  • One extract typically yields a sufficient amount of template for 50–500 PCRs.
  • DNA extracts may be diluted in TE Buffer (1:5) or 10 mM Tris-HCl pH 8–8.5 (1:5–1:10) for long-term storage at -20°C. Extracts are stable at -20°C for ≥6 months.
  • KAPA2G Robust HotStart® ReadyMix is recommended for consistent and reliable endpoint amplification of DNA extracts generated with KAPA Express Extract.
  • No quantification of DNA required prior to PCR.

Quality

All components contained with KAPA Express Extract Kits are subjected to stringent quality control tests, are free of contaminating exo- and endonuclease activities, and meet strict requirements with respect to DNA contamination.

Preparation Note

Handling:
KAPA2G Robust HotStart ReadyMix may not freeze solidly, even when stored at -20°C. This will not affect the shelf-life of the product. Always ensure that the product has been fully thawed and mixed before use.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

FTA is a trademark of Qiagen Group
HOTSTART is a registered trademark of Molecular BioProducts, Inc.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • KAPA Express Extract Enzyme 1 U/µL

  • 10X KAPA Express Extract Buffer

  • 2X KAPA2G Robust HotStart® ReadyMix

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feasibility of non-invasive molecular method for sexing of parrots
Bosnjak J, et al.
Pakistan Journal of Zoology, 45 (2013)
Detection and analysis of new psittacine beak and feather disease virus (PBFDv) nucleotide sequences.
Vucicevic M, et al.
Journal of the Hellenic Veterinary Medical Society, 68(4) (2017)
Efficient genetic manipulation of the NOD-Rag1-/-IL2RgammaC-null mouse by combining in vitro fertilization and CRISPR/Cas9 technology
Feng Li
Scientific Reports, 4 (2014)
Detection and analysis of new psittacine beak and feather disease virus (PBFDv) nucleotide sequences
Vucicevic M, et al.
Journal of the Hellenic Veterinary Medical Society, 68, 653-660 (2017)
Kit Receptor Expression in Canine Cutaneous Mast Cell Tumors (CMCTs) Without C-Kit Mutation.
Vucicevic Ivana, et al.
Acta veterinaria, 66(2), 222-233 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service