Skip to Content
Merck

Skip To

D7440

JumpStart Taq ReadyMix for Quantitative PCR

For probe-based real-time PCR

Synonym(s):

hot start DNA polymerase, hot start PCR, hot start master mix, qPCR, real time quantitative PCR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.55
UNSPSC Code:
41106300
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

usage

sufficient for 100 reactions, sufficient for 20 reactions, sufficient for 400 reactions

feature

dNTPs included, hotstart

concentration

2.5 units/reaction (50 μL reaction volume)

technique(s)

RT-qPCR: suitable, qPCR: suitable, whole genome amplification: suitable

color

colorless

input

purified DNA

application(s)

agriculture

compatibility

Bio-Rad MyiQ ( ), Bio-Rad iCycler iQ, Bio-Rad iQ 5, for use with ABI 5700, for use with ABI 7000, for use with ABI 7300, for use with ABI 7500 Fast, for use with ABI 7500, for use with ABI 7700, for use with ABI 7900 Fast, for use with ABI 7900 HT, for use with ABI 7900, for use with ABI StepOne, for use with ABI StepOnePlus, for use with ABI ViiA 7, for use with Bio-Rad CFX384, for use with Bio-Rad CFX96, for use with Bio-Rad MJ Chromo4, for use with Bio-Rad MJ Opticon 2, for use with Bio-Rad MJ Opticon Cepheid SmartCycler, for use with Bio-Rad MJ Opticon, for use with Bio-Rad MiniOpticon, for use with Eppendorf® Mastercycler ep realplex2 s, for use with Eppendorf® Mastercycler ep realplex, for use with Illumina Eco qPCR, for use with Qiagen Corbett Rotor-Gene 3000, for use with Qiagen Corbett Rotor-Gene 6000, for use with Qiagen Corbett Rotor-Gene Q, for use with Roche LightCycler 480, for use with Strategene Mx3000P, for use with Strategene Mx3005P, for use with Strategene Mx4000

detection method

probe-based

shipped in

wet ice

storage temp.

−20°C

General description

JumpStart Taq ReadyMix is a ready-to-use 2X master mix that contains JumpStart Taq DNA polymerase, 99% pure dNTPs, reaction buffer and JumpStart Taq antibody. It combines the performance enhancements of our JumpStart Taq Antibody for hot start PCR with the convenience of an easy-to-use reaction mixture. Since it has no added dyes, this is the ideal solution for performing high-throughput, quantitative PCR methods that rely on a fluorescent probe. The Taq DNA polymerase is an antibody-inactivated hot-start enzyme. Once the reaction temperature reaches 70°C, the DNA polymerase-antibody complex dissociates and Taq DNA polymerase activity is restored. This antibody-enzyme complex allows for easy and convenient set-up with less contamination risk than manual hot-start techniques.

Application

JumpStart Taq ReadyMix for Quantitative PCR has been used:

  • for quantitative polymerase chain reaction (qPCR) amplification of HIV-1 RNA that is reverse transcribed and DNA using probes
  • in a 2-step RT-qPCR assay for the quantification of reverse transcribed RNA in a study to monitor the activity of licorice in Treg cell differentiation and function
  • as a component of the reaction mixture for the detection and amplification of Salmonella sp. and internal amplification control (IAC) pUC 19 plasmid DNA
  • as a component of the reaction mixture for the detection of Clostridium difficile by quantitative polymerase chain reaction (qPCR)
  • for quantitative real-time -PCR of reverse-transcribed ß-arrestin-1 (ARRB1) and β-arrestin-2 (ARRB2) mRNA

Biochem/physiol Actions

The ReadyMix contains JumpStart Taq DNA Polymerase, 99% pure deoxynucleotides, and buffer in an optimized 2× concentrate. To prepare a reaction, 25 μL of the ReadyMix is added to primers, template, detection chemistry, and water for a total reaction volume of 50 μL. Set up can be performed at room temperature since the JumpStart Taq antibody renders the Taq DNA polymerase inactive. During the first denaturation cycle, the antibody dissociates from the enzyme and full activity is restored. No special preparations or protocol changes are required.

Features and Benefits

  • Ultimate Convenience: ReadyMixes contain all components necessary for QPCR, simply add fluorescent detection chemistry, primers, and template
  • Greater Specificity & Increased Target Yield: JumpStart Taq prevents non-specific amplification and increased target yield
  • Maximum Flexibility: Suitable for use with a variety of detection chemistries including molecular probes and double-stranded binding dyes such as SYBR Green I
  • Designed for use with either plate/tube real-time thermal cyclers or capillary instruments

Packaging

Sigma′s Reference Dye for Quantitative PCR is included separately with this ReadyMix for normalization of the reaction data. The dye has a maximum excitation of 586 nm, and a maximum emission of 605 nm. The instrument settings for ROX reference dye are satisfactory for the measurement of the Reference Dye for Quantitative PCR. A tube of 25 mM MgCl2 is also provided for easy optimization of the qPCR reaction.

Default reaction volume is 50 μL

20RXN is packaged as 1 X 500 μL
100RXN is packaged as 1 X 2.5 mL
400RXN is packaged as 1 X 10 mL

Other Notes

JumpStart Taq ReadyMix for Quantitative PCR combines the advantages of a hot start enzyme with a ready-to-use mix for high throughput, quantitative PCR (qPCR). It is formulated without a detection chemistry making it suitable for use with a variety of formats including dual-labeled probes, molecular beacons, or double stranded binding dyes such as SYBR Green I.
One unit incorporates 10 nmol of total dNTPs into acid-precipitable DNA in 30 min. at 74 °C.

Legal Information

SYBR is a trademark of Thermo Fisher Scientific or its subsidiaries
A license to perform the patented 5′ Nuclease Process for research is obtained by the purchase of (i) both Authorized 5′ Nuclease Core Kit and Licensed Probe, (ii) a Licensed 5′ Nuclease Kit, or (iii) license rights from Applied Biosystems.

This product is an Authorized 5′ Nuclease Core Kit. Use of this product is covered by one or more of the following claims outside the US corresponding to US Patent No. 5,210,015 and 5,487,972. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. Separate purchase of a Licensed Probe would convey rights under US Patents and corresponding patent claims outside the US: 5,538,848, 5,723,591, 5,876,930, 6,030,787, 6,258,569, 5,804,375 (claims 1-12 only), and and claims outside the United States corresponding to US Patent No. 6,214,979. No right under any other patent claim and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained from the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Eppendorf is a registered trademark of Eppendorf SE
JumpStart is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S9194P1107P0982
description

For probe-based real-time PCR

description

SYBR Green qPCR reagent, passive reference dye included

description

for High-throughput PCR of complex templates

description

for PCR

technique(s)

whole genome amplification: suitable, qPCR: suitable, RT-qPCR: suitable

technique(s)

whole genome amplification: suitable, qPCR: suitable

technique(s)

PCR: suitable, whole genome amplification: suitable

technique(s)

PCR: suitable, whole genome amplification: suitable

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

concentration

2.5 units/reaction (50 μL reaction volume)

concentration

1.25 units/reaction (50 μL reaction volume)

concentration

0.75 units/reaction (50 μL reaction volume)

concentration

2.5 units/reaction (50 μL reaction volume)

feature

dNTPs included, hotstart

feature

dNTPs included, hotstart

feature

Difficult Templates/Specialty Enzymes PCR, dNTPs included, hotstart

feature

dNTPs included, hotstart

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

detection method

probe-based

detection method

SYBR Green

detection method

-

detection method

-


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • JumpStart Taq ReadyMix, Complete optimized reagent for hot-start PCR at 2X concentration 2 X
    SDS

  • 25 mM MgCl2 1.5 μL
    SDS

  • Reference Dye for Quantitative PCR, 100 ×, solution 100 X
    SDS

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

Active Filters

  1. 1 Ratings-only review