Skip to Content
Merck
All Photos(5)

Key Documents

KK5121

Roche

KAPA2G Fast Genotyping Mix

suitable for PCR, 2 ×

Synonym(s):

PCR, genotyping

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.54

usage

sufficient for 500 reactions
25 μL sufficient for 500 reactions

Quality Level

shelf life

≤18 mo.

feature

dNTPs included
hotstart: no

manufacturer/tradename

Roche

concentration

2 ×

technique(s)

PCR: suitable
genotyping: suitable

input

crude DNA

storage temp.

−20°C

General description

KAPA2G Fast Genotyping Mix (2X) is a ready-to-use master mix containing all components for fast polymerase chain reaction (PCR), except primers and template. The ReadyMix also contains two inert dyes, allowing for the analysis of PCR reaction products by agarose gel electrophoresis without the need to add a DNA loading solution. The KAPA Express Extract system contains a thermostable protease, designed for rapid sample lysis. The KAPA Express Extract, a novel thermostable protease and buffer system allows the extraction of PCR-ready DNA from mouse tissue in as little as 15 minutes, and KAPA2G Fast Genotyping Mix with dye, containing a DNA polymerase engineered via directed evolution for high processivity and extreme speed. The combination of KAPA Express Extract and KAPA2G Fast Genotyping Mix allows for the reliable extraction and amplification of DNA fragments from mouse tissue in as little as 1 hour, as compared to ≥1 day with conventional protocols.

Application

KAPA2G Fast Genotyping Mix:
  • has been used for the extraction and amplification from mouse tail, ear, and toe tissue
  • has been used for the extraction and amplification of DNA from other animal tissue and hyridomas
  • is suitable for use in polymerase chain reaction (PCR)

Biochem/physiol Actions

DNA extracted with KAPA Express Extract is amplified with KAPA2G Fast Genotyping Mix with dye. DNA fragments generated with KAPA2G Fast Genotyping Mix are 3′-dA-tailed and may be cloned into TA cloning vectors, or used for routine downstream analyses or applications, including restriction enzyme digestion, cloning and sequencing. Like wild-type Taq, KAPA2G Fast has 5′→3′ polymerase and 5′→3′ exonuclease activities, but no 3′→5′ exonuclease (proofreading) activity. The fidelity of KAPA2G Fast is like that of wild-type Taq; it has an error rate of approximately 1 error per 1.7 x 105 nucleotides incorporated. Lysis is performed in a thermocycler, heating block or water bath, after which the sample is centrifuged to recover the DNA containing supernatant. The supernatant is then diluted 10-fold before use as a template in PCR. Unlike existing protocols that rely on Proteinase K digestion, extractions using KAPA Express Extract are conveniently performed in a single tube, without the need for hazardous chemicals, or multiple wash steps. This greatly reduces the risk of sample loss and contamination. The process yields enough template for multiple assays and is easily scaled to handle samples in 96-well format. Each extraction yields enough template for at least 100 genotyping PCR reactions. Extracts (either diluted or undiluted) are stable at -15°C to -25°C for at least 6 months.

Features and Benefits

Increased throughput, turnaround time and reliability:
  • PCR-ready DNA generated in as little as 15 minutes
  • Minimal handling, thereby reducing the risk of sample loss or contamination
  • Sufficient template for multiple assays; easily scaled to handle samples in a 96-well format
  • Fast and reliable amplification of DNA fragments across a wide range of amplicon lengths and GC contents

Outperforms crude extraction methods and commercial kits:
  • Increased turnaround times
  • Increased specificity
  • Decreased cost

Quick Notes:
  • Extract PCR-ready DNA from mouse tail, ear, or toe tissue in 15 minutes.Dilute DNA extract 10-fold prior to PCR in 10 mM Tris-HCl (pH 8.0–8.5), and use 1 μL of DNA per 25 μL reaction.
  • Extracts are stable at -20°C for >6 months.
  • KAPA2G Fast (HotStart) Genotyping Mix contains the engineered KAPA2G Fast DNA Polymerase for fast PCR, along with buffer, MgCl2 (1.5 mM at 1X), and dNTPs (0.2 mM each at 1X).
  • Loading dye for electrophoresis is included in the KAPA2G Fast Genotyping Mix, which allows direct loading of PCR products onto agarose gels for analysis.
  • Use 15 sec annealing time, with 15 sec extension time for amplicons <1 kb, or 15–30 sec/kb extension for longer amplicons.

Quality

All components contained with KAPA Mouse Genotyping Kits are subjected to stringent quality control tests, are free of contaminating exo- and endonuclease activities and meet strict requirements with respect to DNA contamination.

Preparation Note

Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for long-term storage. Provided that all components have been handled carefully and not contaminated, the kit is not expected to be compromised if left (unintentionally) at room temperature for a short period of time (up to 3 days).

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • KAPA2G Fast DNA Polymerase

  • KAPA2G Buffer A

  • dNTPs 0.2 mM each at 1X

  • MgCl2 1.5 mM at 1X

  • stabilizers

  • dye

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunogenomic engineering of a plug-and-(dis) play hybridoma platform.
Pogson M, et al.
Nature Communications, 7, 12535-12535 (2016)
Veroniqa Lundbäck et al.
Physiological reports, 8(16), e14538-e14538 (2020-08-20)
In obesity, the expression level of thyroid stimulating hormone receptor in adipose tissue is reduced and the levels of thyroid stimulating hormone (TSH) are often elevated within the normal range. To investigate the role of TSHR in brown and white

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service