Skip to Content
Merck
All Photos(5)

Documents

MGKITKB

Roche

KAPA Mouse Genotyping Kit

Synonym(s):

genotyping

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

usage

sufficient for 100 reactions (KK7301)
sufficient for 500 reactions (KK7302)

Quality Level

shelf life

≤18 mo.

feature

dNTPs included
hotstart: no

manufacturer/tradename

Roche

technique(s)

PCR: suitable
genotyping: suitable

input

crude DNA

storage temp.

−20°C

General description

KAPA Mouse Genotyping Kits include KAPA Express Extract, a novel thermostable protease and buffer system that allows for the extraction of PCR-ready DNA from mouse tissue in as little as 15 minutes, and KAPA2G Fast Genotyping Mix with dye, containing a DNA polymerase engineered via directed evolution for high processivity and extreme speed. The combination of KAPA Express Extract and KAPA2G Fast Genotyping Mix allows for the reliable extraction and amplification of DNA fragments from mouse tissue in as little as 1 hour, as compared to ≥1 day with conventional protocols.KAPA2G Fast Genotyping Mix (2X) is a ready-to-use master mix containing all components for fast PCR, except primers and templates.

Application

KAPA Mouse Genotyping Kit has been used in the DNA extraction and amplification from mouse tail, ear, and toe tissue. The kit is also suitable for extraction and amplification of DNA from other animal tissue, but protocol optimization may be required.

Biochem/physiol Actions

DNA fragments generated with KAPA2G Fast Genotyping Mix are 3′-dA-tailed and may be cloned into TA cloning vectors, or used for routine downstream analyses or applications, including restriction enzyme digestion, cloning, and sequencing. Like wild-type Taq, KAPA2G Fast has 5′→3′ polymerase and 5′→3′ exonuclease activities, but no 3′→5′ exonuclease (proofreading) activity. The fidelity of KAPA2G Fast is similar to that of wild-type Taq; it has an error rate of approximately 1 error per 1.7 x 105 nucleotides incorporated.

Features and Benefits

Increased throughput, turnaround time and reliability:
  • PCR-ready DNA generated in as little as 15 minutes
  • Minimal handling, thereby reducing the risk of sample loss or contamination
  • Sufficient template for multiple assays; easily scaled to handle samples in a 96-well format
  • Fast and reliable amplification of DNA fragments across a wide range of amplicon lengths and GC contents

Outperforms crude extraction methods and commercial kits:
  • Increased turnaround times Increased specificity Decreased cost

Quick Notes:
  • Extract PCR-ready DNA from mouse tail, ear, or toe tissue in 15 minutes.
  • Dilute DNA extract 10-fold prior to PCR in 10 mM Tris-HCl (pH 8.0–8.5), and use 1 μL of DNA per 25 μL reaction.
  • Extracts are stable at -20°C for >6 months.
  • KAPA2G Fast (HotStart) Genotyping Mix contains the engineered KAPA2G Fast DNA Polymerase for fast PCR, along with buffer, MgCl2 (1.5 mM at 1X), and dNTPs (0.2 mM each at 1X).
  • Loading dye for electrophoresis is included in the KAPA2G Fast Genotyping Mix, which allows direct loading of PCR products onto agarose gels for analysis.
  • Use 15 sec annealing time, with 15 sec extension time for amplicons <1 kb, or 15–30 sec/kb extension for longer amplicons.

Quality

All components contained with KAPA Mouse Genotyping Kits are subjected to stringent quality control tests, are free of contaminating exo- and endonuclease activities and meet strict requirements with respect to DNA contamination.

Preparation Note

Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for long-term storage. Provided that all components have been handled carefully and not contaminated, the kit is not expected to be compromised if left (unintentionally) at room temperature for a short period of time (up to 3 days).

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • 2X KAPA2G Fast Standard or HotStart® Genotyping Mix with dye

  • KAPA Express Extract 1 U/μL

  • 10X KAPA Express Extract Buffer

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siavash Ghaffari et al.
Arteriosclerosis, thrombosis, and vascular biology, 41(1), 200-216 (2020-10-16)
LDL (low-density lipoprotein) transcytosis across the endothelium is performed by the SR-BI (scavenger receptor class B type 1) receptor and contributes to atherosclerosis. HMGB1 (high mobility group box 1) is a structural protein in the nucleus that is released by
Zhimiao Lin et al.
Nature genetics, 48(12), 1508-1516 (2016-11-01)
Skin integrity is essential for protection from external stress and trauma. Defects in structural proteins such as keratins cause skin fragility, epitomized by epidermolysis bullosa (EB), a life-threatening disorder. Here we show that dominant mutations of KLHL24, encoding a cullin
Stéphane Dalle et al.
Methods in molecular biology (Clifton, N.J.), 1957, 345-364 (2019-03-29)
Novel findings reveal important functional roles for β-arrestin 1 and β-arrestin 2 in the regulation of insulin secretion, β-cell survival, and β-cell mass plasticity not only by glucose but also by G-protein-coupled receptors, such as the glucagon-like peptide-1 (GLP-1) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service