Skip to Content
Merck
All Photos(4)

Documents

2GFHSKB

Roche

KAPA2G Fast HotStart PCR Kit

Mg and Mg-free buffers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

usage

sufficient for 1000 reactions
sufficient for 200 reactions
sufficient for 500 reactions
sufficient for 5000 reactions

shelf life

≤18 mo.

feature

Fast PCR
dNTPs included: no
hotstart

packaging

pkg of 100 U (KK5523)
pkg of 250 U (KK5503)
pkg of 250 U (KK5513; Mg Free)
pkg of 2500 U (KK5519)
pkg of 500 U (KK5501)
pkg of 500 U (KK5511; Mg Free)

manufacturer/tradename

Roche

technique(s)

PCR: suitable

input

purified DNA

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

KAPA2G Fast DNA Polymerase is a second-generation (2G) enzyme engineered for higher processivity and speed, offering significantly faster extension rates than wild-type Taq polymerase. In addition to speed, KAPA2G Fast provides higher yields and sensitivity than competitor enzymes across a broad range of targets. KAPA2G Fast HotStart® PCR Kits are designed for fast PCR, in which total reaction times are 20–70% shorter than those of conventional PCR assays performed with wild-type Taq DNA polymerase. This can be achieved without sacrificing reaction performance, and does not require specialized PCR consumables or thermocyclers.

Application

KAPA2G Fast HotStart PCR Kit has been used for:
  • Fast PCR
  • Genotyping
  • The quantification of chromatin immunoprecipitation (ChIP)-seq libraries
  • Routine polymerase chain reaction (PCR)

Biochem/physiol Actions

KAPA2G Fast DNA Polymerase generates DNA fragments, which share the same characteristics as DNA fragments generated with wild-type Taq DNA polymerase. It is useful for routine downstream analyses, like restriction enzyme digestion, cloning and sequencing. Similar to wild-type Taq, KAPA2G Fast has 5′→3′ polymerase and 5′→3′ exonuclease activities. But it has no 3′→5′ exonuclease (proofreading) activity. The fidelity of KAPA2G Fast is comparable to wild-type Taq, which has an error rate of approximately 1 error per 1.7 x 105 nucleotides incorporated.

Features and Benefits

Reduce cycling times up to 75% :
  • Extension times as low as 1 sec per kilobase
  • Broad coverage of both AT- and GC-rich targets

High speed, higher performance :
  • Single copy sensitivity from complex targets
  • Increased yields

Quick Notes :

  • KAPA2G Fast HotStart® PCR Kits contain the engineered KAPA2G Fast HotStart DNA Polymerase, developed for fast PCR.
  • Use 1 sec extension time for amplicons <1 kb and 15 sec/kb for longer amplicons, and save 20–70% of total reaction time.
  • No need for specialized instrumentation or PCR consumables.
  • Optimized buffer system offers improved yields,specificity and sensitivity, facilitating efficient primer annealing across a wide range of primer lengths,GC contents, and melting temperatures.
  • Use 0.5 U KAPA2G Fast HotStart DNA Polymerase per 25 μL reaction, or less for smaller volumes.
  • For high reaction efficiency, do not exceed 25 μL reaction volumes.
  • The buffer is available with and without MgCl2 KAPA2G Buffer A contains 1.5 mM MgCl2 at 1X, while KAPA2G Buffer M is Mg-free.Aside from MgCl2 content, these two buffers are identical.
  • Reaction products are 3′-dA-tailed and may be cloned into TA cloning vectors.

Quality

Each batch of KAPA2G Fast DNA Polymerase is confirmed to contain <2% contaminating protein (Agilent Protein 230 Assay). KAPA2G Fast ReadyMix is subjected to stringent quality control tests, is free of contaminating exo- and endonuclease activity, and meets strict requirements with respect to DNA contamination levels.

Preparation Note

Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for long-term storage. Provided that all components have been handled carefully and not contaminated, the kit is not expected to be compromised if left (unintentionally) at room temperature for a short period of time (up to 3 days). Long-term storage at room temperature and 4°C is not recommended. Please note that reagents stored at temperatures above -20°C are more prone to degradation when contaminated during use, and therefore storage at such temperatures is at the user′s own risk.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

HOTSTART is a registered trademark of Molecular BioProducts, Inc.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • KAPA2G Fast HotStart® DNA Polymerase 5 U/µL

  • 5X KAPA2G Buffer A

  • 5X KAPA2G Buffer M (Mg-free)

  • KAPA MgCl2 25 mM

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - STOT SE 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Express barcodes: racing from specimen to identification.
Ivanova N V, et al.
Molecular Ecology Resources, 9, 35-41 (2009)
Paola Brescia et al.
Cancer cell, 34(3), 453-465 (2018-09-12)
The gene encoding the MEF2B transcription factor is mutated in germinal center (GC)-derived B cell lymphomas, but its role in GC development and lymphomagenesis is unknown. We demonstrate that Mef2b deletion reduces GC formation in mice and identify MEF2B transcriptional
Francesca Del Bufalo et al.
Frontiers in oncology, 8, 526-526 (2018-11-30)
Low-grade gliomas (LGG) are the most common central nervous system tumors in children. Prognosis depends on complete surgical resection. For patients not amenable of gross total resection (GTR) new approaches are needed. The BRAF mutation V600E is critical for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service