Skip to Content
Merck
All Photos(1)

Key Documents

10108090001

Roche

Peroxidase (POD)

grade I, from horseradish

Synonym(s):

POD, peroxidase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

biological source

horseradish

Quality Level

form

lyophilized (salt-free)

specific activity

≥250 units/mg protein (At 25 °C with guaiacol and H2O2 as the substrates.)
250 U/mg

packaging

pkg of 25,000 U

manufacturer/tradename

Roche

optimum pH

6.0-6.5

shipped in

wet ice

storage temp.

2-8°C

General description

Donor:hydrogen-peroxide oxidoreductase
Peroxidase (POD) (E.C. 1.11.1.7) is extensively expressed in most fruits and vegetables.[1]

Application

Oxidation of electron donors (e.g., epoxidation of double bonds). Peroxidase can be coupled to other proteins via its amino groups, as well as its carbohydrate moiety. The reconstituted solution can be used directly for conjugation without prior dialysis.It has been used in ligand blotting.[2]
Peroxidase (POD) has been used in AUR (Amplex UltraRed assay) to measure extracellular H2O2 produced from disaggregated slug cells.[3]

Biochem/physiol Actions

Peroxidase (POD) is involved in the dehydrogenation of a large number of organic compounds including, phenols and aromatic amines, hydroquinones and hydroquinoid amines, mainly benzidine derivatives.[4]

Quality

Contaminants: <0.001% ATPase and acid phosphatase each, <0.7% catalase
Purity number: approximately 3.0 (A403/A275)

Preparation Note

Activator: Ammonia, pyridine, imidazole, all at pH >7; detergents.
Working solution: PBS (phosphate buffered saline) or water (starting concentration 20 mg/ml).

Reconstitution

Solution (10 mg/ml in water): clear, red brown; pH-value: 6.0 to 7.0.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service