Skip to Content
Merck
All Photos(2)

Key Documents

CC130

Sigma-Aldrich

Human Vitronectin

recombinant, expressed in E. coli, liquid, 0.5 mg/mL, used as an effective xeno-free alternative to matrigel, suitable for human ES and iPS cell culture, PLURISTEM®

Synonym(s):

S-protein, Serum-spreading factor, V75

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

product name

PluriSTEM-XF® Recombinant Vitronectin, Human pluripotent stem cell culture.

biological source

Escherichia coli
human

Quality Level

Assay

≥92% (SDS-PAGE)

form

liquid

manufacturer/tradename

PluriSTEM®

concentration

0.5 mg/mL

technique(s)

cell attachment: suitable
cell culture | mammalian: suitable
cell culture | stem cell: suitable

input

sample type: human embryonic stem cell(s)

UniProt accession no.

shipped in

dry ice

General description

PluriSTEM Recombinant Vitronectin is an effective xeno-free alternative to matrigel and has been validated to successully culture human ES and iPS cells using PluriSTEM Human ES/iPS Medium (Millipore Cat. No. SCM130, SCM132), E8 and NutriStem human ES cell growth media.

PluriSTEM Recombinant Vitronectin is a truncated form of human vitronectin (gene 20-398 aa fragment) that was constructed with codon optimization, expressed in E. coli and purified as inclusing bodies. The final product was refolded using a unique “temperature shift inclusion body refolding” technology and chromatographically purified. The final product is considered xeno-free.

Application

Research Category
Stem Cell Research

Cell Structure

ECM & Adhesion
Supports feeder-free and serum-free growth of human es/ips cells while maintaining xeno-free culture conditions.

Quality

Each lot is tested on human ES cells (H1) cultured using 5-10 μg recombinant vitronectin in 1 mL human ESC medium per well (6-well plate).

Physical form

Product is sterile-filtered in a 20 mM pH 8.0 Tris-HCl buffer, with a proprietary formulation of NaCl, KCl, EDTA, arginine, DTT and glycerol.

Storage and Stability

Store at -20°C until expiration date on vial. Once thawed, product is stable at 2 - 8°C for two weeks. Thawed vitronectin can be aliquoted and stored at -20°C. Avoid additional freeze-thaw cycles.

Legal Information

PLURISTEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutsumi Kobayashi et al.
Stem cell reports, 17(3), 507-521 (2022-02-13)
In vitro expansion of human primordial germ cell-like cells (hPGCLCs), a pluripotent stem cell-derived PGC model, has proved challenging due to rapid loss of primordial germ cell (PGC)-like identity and limited cell survival/proliferation. Here, we describe long-term culture hPGCLCs (LTC-hPGCLCs), which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service