Skip to Content
Merck
All Photos(1)

Documents

08-126

Sigma-Aldrich

Human Vitronectin

recombinant, expressed in E. coli, liquid, 1.5 mg/mL, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

product name

Vitronectin, human recombinant,

biological source

human

Quality Level

recombinant

expressed in E. coli

form

liquid

mol wt

Mw 78 kDa

manufacturer/tradename

Upstate®

concentration

1.5 mg/mL

technique(s)

cell culture | mammalian: suitable

input

sample type hematopoietic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type neural stem cell(s)
sample type: human embryonic stem cell(s)
sample type mesenchymal stem cell(s)
sample type epithelial cells
sample type pancreatic stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... VTN(7448)

General description

Product Source: N-terminal, GST-tagged, full-length human Vitronectin expressed in E. coli

Quality

Routinely evaluated by cell adhesion assay.

Linkage

Replaces: CC080

Physical form

Liquid in 10mM Sodium Phosphate, pH 7.1, 150mM Sodium Chloride, 0.1mM DTT, 0.1mM PMSF and 50% glycerol.

Storage and Stability

6 months at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression, production, and characterization of full-length vitronectin in Escherichia coli
Wojciechowski, Katherine, et al
Protein Expression and Purification, 36, 131-138 (2004)
R J Wechsler-Reya
Trends in neurosciences, 24(12), 680-682 (2001-11-24)
Cerebellar granule cells are the most abundant neurons in the brain and are crucial to the circuitry that controls motor coordination. The proliferation of granule cell precursors (GCPs) is controlled by the secreted signaling molecule Sonic hedgehog (Shh), but the
I Schvartz et al.
The international journal of biochemistry & cell biology, 31(5), 539-544 (1999-07-10)
Vitronectin is a multifunctional glycoprotein present in blood and in the extracellular matrix. It binds glycosaminoglycans, collagen, plasminogen and the urokinase-receptor, and also stabilizes the inhibitory conformation of plasminogen activation inhibitor-1. By its localization in the extracellular matrix and its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service